Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00182570_m1
          See other TNFRSF10C GE Assays ›
          Gene Symbol:
          TNFRSF10C
          Gene Name:
          TNF receptor superfamily member 10c
          Gene Aliases:
          CD263, DCR1, DCR1-TNFR, LIT, TRAIL-R3, TRAILR3, TRID
          Chromosome Location:
          Chr.8: 23102814 - 23117437 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00182570_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TNFRSF10C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003841.3 NP_003832.2 4 - 5 697 81
          GenBank mRNA AF012536.1 - 4 - 5 581 81
          AF012629.1 - 4 - 5 389 81
          AF014794.1 - 4 - 5 538 81
          AF020502.1 - 4 - 5 509 81
          AF033854.1 - 4 - 5 565 81
          AK298664.1 - 3 - 4 484 81
          AK313213.1 - 4 - 5 591 81
          AY358281.1 - 4 - 5 581 81
          BC017852.1 - 4 - 5 590 81
          BC125041.1 - 4 - 5 510 40115748 81
          BC125042.1 - 4 - 5 510 40115749 81
          CR541857.1 - 4 - 5 389 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800007007.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000180.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Recommended Assay Homo sapiens Human TNF Superfamily Pathway HUMAN APOPTOSIS THROUGH DEATH RECEPTORS 1700 Inventoried Human_TNF_Superfamily_Pathway Human_Apoptosis_through_Death_Receptors Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TNFRSF10C

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          immune response
          signal transduction
          multicellular organism development
          extrinsic apoptotic signaling pathway via death domain receptors
          response to lipopolysaccharide
          tumor necrosis factor-mediated signaling pathway
          regulation of cell proliferation
          regulation of apoptotic process
          transmembrane signaling receptor activity
          tumor necrosis factor-activated receptor activity
          TRAIL binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000518135.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000518135.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000518135.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000518135.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000518135.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000520607.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000520607.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000520607.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000520607.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000520607.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 ENST00000520607.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08002342.hg.1 TC08000180.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08002342.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08002342.hg.1 ENST00000540813.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08002342.hg.1 ENST00000544885.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08002342.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000397703.6 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000518135.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000518135.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000518135.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000518135.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000518135.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000520607.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000520607.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000520607.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000520607.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000520607.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 ENST00000520607.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007007.hg.1 NR_046173.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800147951.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800147951.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800147951.hg.1 ENST00000518135.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800147951.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800147951.hg.1 ENST00000356864.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800147951.hg.1 ENST00000517558.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800147951.hg.1 ENST00000518135.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800147951.hg.1 NM_003841.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800147951.hg.1 TC0800007007.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 138830 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline