Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00184921_m1
          See other ASNA1 GE Assays ›
          Gene Symbol:
          ASNA1
          Gene Name:
          arsA arsenite transporter, ATP-binding, homolog 1 (bacterial)
          Gene Aliases:
          ARSA-I, ARSA1, ASNA-I, GET3, TRC40
          Chromosome Location:
          Chr.19: 12737492 - 12748323 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00184921_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ASNA1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF047469.1 - 1 - 2 167 93
          AK223363.1 - 1 - 2 174 93
          AK291855.1 - 1 - 2 168 93
          BC002651.2 - 1 - 2 160 3609717 93
          BG426136.1 - 1 - 2 174 93
          BM718941.1 - 1 - 2 215 93
          U60276.1 - 1 - 2 111 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900007089.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transporter

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100010719.hg.1 ENST00000603591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010719.hg.1 ENST00000603591.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150933.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150933.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150933.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150933.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150933.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900150933.hg.1 TC1900007089.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900150933.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150933.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150933.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150933.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150933.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 TC1900007089.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150935.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 TC1900007089.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900150934.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000357332.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000586561.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000590633.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000591090.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 ENST00000611688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
          Affymetrix Human Clariom D Assay TC1900007089.hg.1 NM_004317.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 121011 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0