Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00204257_m1
          See other CD274 GE Assays ›
          Gene Symbol:
          CD274
          Gene Name:
          CD274 molecule
          Gene Aliases:
          B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1, hPD-L1
          Chromosome Location:
          Chr.9: 5450542 - 5470554 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00204257_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CD274
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001314029.2 NP_001300958.1 3 - 4 464 77
          NM_014143.4 NP_054862.1 3 - 4 464 77
          NR_052005.2 3 - 4 464 77
          XM_047423262.1 XP_047279218.1 2 - 3 398 77
          GenBank mRNA AA399416.1 - 1 - 2 405 77
          AF177937.1 - 2 - 3 395 77
          AF233516.1 - 3 - 4 447 77
          AK001894.1 - 1 - 2 210 77
          AK300470.1 - 3 - 4 464 77
          AK314567.1 - 3 - 4 464 77
          AY254342.1 - 2 - 3 395 77
          AY714881.1 - 2 - 3 395 77
          BC069381.1 - 3 - 4 445 77
          BC074984.2 - 2 - 3 415 77
          BC113734.1 - 2 - 3 416 77
          BC113736.1 - 2 - 3 416 77
          DB160805.1 - 3 - 4 434 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900006559.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000037.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Immune Tolerance Network Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CD274

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          immune system process
          negative regulation of T cell mediated immune response to tumor cell
          immune response
          signal transduction
          cell surface receptor signaling pathway
          T cell costimulation
          negative regulation of interferon-gamma production
          negative regulation of interleukin-10 production
          positive regulation of interleukin-10 production
          response to cytokine
          TRIF-dependent toll-like receptor signaling pathway
          positive regulation of T cell proliferation
          negative regulation of T cell proliferation
          positive regulation of transcription, DNA-templated
          regulation of activated T cell proliferation
          negative regulation of activated T cell proliferation
          negative regulation of T cell activation
          regulation of T cell apoptotic process
          cellular response to lipopolysaccharide
          negative regulation of tumor necrosis factor superfamily cytokine production
          regulation of activated CD4-positive, alpha-beta T cell apoptotic process
          positive regulation of activated CD8-positive, alpha-beta T cell apoptotic process
          negative regulation of CD4-positive, alpha-beta T cell proliferation
          negative regulation of CD8-positive, alpha-beta T cell activation
          transcription coactivator activity
          protein binding
          receptor agonist activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381573.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000381577.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000474218.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000474218.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000474218.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000474218.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000474218.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000474218.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000492923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000492923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000492923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000492923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000492923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000492923.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000498261.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000498261.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000498261.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000498261.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000498261.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000498261.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000498261.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 ENST00000498261.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_001267706.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NM_014143.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006559.hg.1 NR_052005.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 208063 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline