Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00216479_m1
          See other CDCA8 GE Assays ›
          Gene Symbol:
          CDCA8
          Gene Name:
          cell division cycle associated 8
          Gene Aliases:
          BOR, BOREALIN, DasraB, MESRGP
          Chromosome Location:
          Chr.1: 37692401 - 37709719 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00216479_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CDCA8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001256875.1 NP_001243804.1 4 - 5 628 127
          NM_018101.3 NP_060571.1 5 - 6 531 127
          GenBank mRNA AK001330.1 - 5 - 6 451 127
          AK022104.1 - 4 - 5 533 127
          AK022606.1 - 4 - 5 437 127
          AK222549.1 - 5 - 6 451 127
          AK222568.1 - 5 - 6 405 127
          AK291917.1 - 4 - 5 483 127
          AK302116.1 - 3 - 4 295 127
          AY508815.2 - 5 - 6 451 127
          BC000703.2 - 4 - 5 524 3347875 127
          BC001651.2 - 4 - 5 542 2822261 127
          BC008079.2 - 5 - 6 384 2959649 127
          BC016944.1 - 5 - 6 404 3457122 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100007841.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000476.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CDCA8

          Gene Ontology Categories:

          Function(s) Process(es)

          sister chromatid cohesion
          mitotic metaphase plate congression
          protein sumoylation
          chromosome organization
          cell division
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01007408.hg.1 TC01000476.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01007408.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007408.hg.1 ENST00000373055.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01007408.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100155222.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155222.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155222.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100155222.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100155222.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155222.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100155222.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100155222.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100155222.hg.1 TC0100007841.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000327331.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 ENST00000373055.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_001256875.1 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq
          Affymetrix Human Clariom D Assay TC0100007841.hg.1 NM_018101.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline