Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00218350_m1
          See other FBXO6 GE Assays ›
          Gene Symbol:
          FBXO6
          Gene Name:
          F-box protein 6
          Gene Aliases:
          FBG2, FBS2, FBX6, Fbx6b
          Chromosome Location:
          Chr.1: 11664093 - 11674354 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00218350_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FBXO6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018438.5 NP_060908.1 4 - 5 673 63
          XM_005263448.4 XP_005263505.1 4 - 5 613 63
          XM_005263449.4 XP_005263506.1 4 - 5 560 63
          XM_005263451.4 XP_005263508.1 4 - 5 613 63
          XM_006710571.3 XP_006710634.1 4 - 5 641 63
          XM_011541232.2 XP_011539534.1 4 - 5 607 63
          XM_017001007.1 XP_016856496.1 5 - 6 675 63
          GenBank mRNA AF129536.1 - 4 - 5 642 63
          AF233223.1 - 3 - 4 518 63
          AK314989.1 - 4 - 5 673 63
          BC020880.1 - 4 - 5 615 4614957 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100006862.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000145.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FBXO6

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint
          protein polyubiquitination
          DNA repair
          proteolysis
          glycoprotein catabolic process
          response to unfolded protein
          ER-associated ubiquitin-dependent protein catabolic process
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          glycoprotein binding
          ubiquitin-protein transferase activity
          protein binding
          carbohydrate binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01002281.hg.1 TC01000145.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01002281.hg.1 ENST00000376753.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01002281.hg.1 NM_018438.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000449067.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000449067.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000449067.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000449067.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000449067.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000474239.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000474239.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000474239.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000474239.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000474239.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 ENST00000474239.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000449067.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000449067.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000449067.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000449067.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000449067.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000474239.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000474239.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000474239.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000474239.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000474239.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 ENST00000474239.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay TC0100006862.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100147610.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100147610.hg.1 ENST00000449067.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100147610.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100147610.hg.1 ENST00000376753.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0100147610.hg.1 ENST00000449067.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100147610.hg.1 NM_018438.5 Refseq
          Affymetrix Human Clariom D Assay PSR0100147610.hg.1 TC0100006862.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline