Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00235001_m1
          See other NR4A3 GE Assays ›
          Gene Symbol:
          NR4A3
          Gene Name:
          nuclear receptor subfamily 4 group A member 3
          Gene Aliases:
          CHN, CSMF, MINOR, NOR1, TEC
          Chromosome Location:
          Chr.9: 99821855 - 99866893 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00235001_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NR4A3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006981.3 NP_008912.2 6 - 7 2183 107
          NM_173200.2 NP_775292.1 7 - 8 2255 107
          XM_017015162.1 XP_016870651.1 5 - 6 2115 107
          GenBank mRNA AK226108.1 - 6 - 7 2165 107
          D78579.1 - 6 - 7 2185 107
          S81243.1 - 6 - 7 2070 107
          U12767.1 - 5 - 6 1546 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900008219.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000508.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Nuclear Receptor Immune Tolerance Network Homo sapiens Human Tumor Metastasis Panel Inventoried Human Tumor Metastasis Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NR4A3

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          mesoderm formation
          transcription initiation from RNA polymerase II promoter
          gastrulation
          axon guidance
          biological_process
          pyruvate oxidation
          positive regulation of cardiac muscle hypertrophy
          positive regulation of glucose transport
          hippocampus development
          intracellular receptor signaling pathway
          adult behavior
          organ regeneration
          positive regulation of mast cell cytokine production
          common myeloid progenitor cell proliferation
          positive regulation of mast cell activation by Fc-epsilon receptor signaling pathway
          response to hydrogen peroxide
          mast cell degranulation
          steroid hormone mediated signaling pathway
          negative regulation of neuron apoptotic process
          cellular response to leptin stimulus
          cellular respiration
          fat cell differentiation
          positive regulation of cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fatty acid oxidation
          regulation of smooth muscle cell proliferation
          positive regulation of smooth muscle cell proliferation
          semicircular canal morphogenesis
          positive regulation of epithelial cell proliferation
          neuromuscular process controlling balance
          vestibular reflex
          regulation of type B pancreatic cell proliferation
          cellular response to corticotropin-releasing hormone stimulus
          cellular response to catecholamine stimulus
          energy homeostasis
          positive regulation of monocyte aggregation
          negative regulation of hydrogen peroxide-induced neuron death
          positive regulation of leukocyte apoptotic process
          positive regulation of feeding behavior
          regulation of energy homeostasis
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription coactivator binding
          DNA binding
          steroid hormone receptor activity
          thyroid hormone receptor activity
          protein binding
          zinc ion binding
          protein kinase binding
          histone acetyltransferase binding
          glucocorticoid receptor binding
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09005521.hg.1 TC09000508.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09005521.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005521.hg.1 ENST00000395097.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09005521.hg.1 NM_006981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09005521.hg.1 NM_173200.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000338488.8 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_006981.3 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173199.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay TC0900008219.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900155065.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900155065.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900155065.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900155065.hg.1 NM_006981.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900155065.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900155065.hg.1 ENST00000330847.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900155065.hg.1 ENST00000395097.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900155065.hg.1 ENST00000618101.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900155065.hg.1 NM_006981.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900155065.hg.1 NM_173200.2 Refseq
          Affymetrix Human Clariom D Assay PSR0900155065.hg.1 TC0900008219.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0