Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00237013_m1
          See other CCL11 GE Assays ›
          Gene Symbol:
          CCL11
          Gene Name:
          C-C motif chemokine ligand 11
          Gene Aliases:
          SCYA11
          Chromosome Location:
          Chr.17: 34285668 - 34288180 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00237013_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCL11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002986.2 NP_002977.1 1 - 2 219 76
          GenBank mRNA BC017850.1 - 1 - 2 145 4618679 76
          BF197516.1 - 2 - 3 76
          CR457421.1 - 1 - 2 78 76
          U46573.1 - 1 - 2 131 76
          Z75668.1 - 2 - 3 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700007559.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000385.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Immune Tolerance Network Mammalian Gene Collection Recommended Assay Homo sapiens Human Chemokine Panel Inventoried Human_Chemokine Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCL11

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          chronic inflammatory response
          monocyte chemotaxis
          mast cell chemotaxis
          protein phosphorylation
          cellular calcium ion homeostasis
          chemotaxis
          inflammatory response
          cytoskeleton organization
          actin filament organization
          cell adhesion
          signal transduction
          G-protein coupled receptor signaling pathway
          regulation of cell shape
          response to radiation
          response to virus
          positive regulation of cell migration
          neutrophil chemotaxis
          positive regulation of actin filament polymerization
          response to interleukin-13
          positive regulation of GTPase activity
          positive regulation of angiogenesis
          eosinophil chemotaxis
          lymphocyte chemotaxis
          positive regulation of inflammatory response
          branching involved in mammary gland duct morphogenesis
          mammary duct terminal end bud growth
          chemokine-mediated signaling pathway
          ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          response to interleukin-4
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          protein binding
          chemokine activity
          CCR chemokine receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 NM_002986.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 NM_002986.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 NM_002986.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 NM_002986.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 NM_002986.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007559.hg.1 NM_002986.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17004882.hg.1 ENST00000305869.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17004882.hg.1 NM_002986.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17004882.hg.1 TC17000385.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17004881.hg.1 ENST00000305869.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17004881.hg.1 NM_002986.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17004881.hg.1 TC17000385.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17004880.hg.1 NM_002986.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17004880.hg.1 TC17000385.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700154393.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154393.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154393.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154393.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154393.hg.1 TC1700007559.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700154396.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154396.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154396.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154396.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154396.hg.1 TC1700007559.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700154394.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154394.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154394.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154394.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154394.hg.1 TC1700007559.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay TC1700007559.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154391.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154391.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154391.hg.1 ENST00000305869.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700154391.hg.1 NM_002986.2 Refseq
          Affymetrix Human Clariom D Assay PSR1700154391.hg.1 TC1700007559.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline