Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00258453_s1
          See other LINC00597 GE Assays ›
          Gene Symbol:
          LINC00597
          Gene Name:
          long intergenic non-protein coding RNA 597
          Gene Aliases:
          C15orf5
          Chromosome Location:
          Chr.15: 77223893 - 77225404 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00258453_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00597
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026813.1 1 747 76
          GenBank mRNA AF216224.1 - 1 753 76
          AL109679.1 - 1 392 76
          BC069765.1 - 1 683 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500010110.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15001692.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P54447 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P54447 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 11360 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 59356 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P54447 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P54447 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266264 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266222 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770334 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770303 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266249 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266251 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770331 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770312 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266221 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266246 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266222 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770334 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770346 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770331 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266221 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770312 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266229 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266264 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266246 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266251 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09770303 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30266249 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P54447 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 58772 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 101165 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770337 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770351 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770342 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770353 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770308 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770305 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770313 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770320 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770324 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770340 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770335 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770318 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770325 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770323 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770329 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770332 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770327 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770344 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770309 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770316 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770341 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770331 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770311 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770355 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09770347 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7990640 - -
          Affymetrix Human Gene 1.0 ST Array 7990642 - -
          Affymetrix Human Gene 1.0 ST Array 7990639 - -
          Affymetrix Human Gene 1.0 ST Array 7990638 - -
          Affymetrix Human Gene 1.0 ST Array 7990641 - -
          Affymetrix Human Gene 1.0 ST Array 7990637 - -
          Affymetrix Human Gene 1.1 ST Array 7990636 - -
          Affymetrix HT HG-U133+ PM Array Plate 208109_PM_s_at - -
          Affymetrix Human Genome U219 Array 11745030_at - -
          Affymetrix Human Genome U219 Array 11745030_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR15016487.hg.1 NR_026813.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15016487.hg.1 TC15001692.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1500175771.hg.1 NR_026813.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500175771.hg.1 NR_026813.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500175771.hg.1 TC1500010110.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 208109_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 208109_s_at - -
          Human Genome U133A 2.0 Array 208109_s_at - -
          Human Genome U133A 2.0 Array 208109_PM_s_at - -
          AffymetrixPathwork Diagnostics Pathchip pw584000 - -
          Human Genome U95 Set 34140_at - -
          Human Genome U95 Set 34140_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208109_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208109_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208109_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208109_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208109_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208109_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208109_s_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208109_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208109_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208109_s_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208109_s_at11 - -
          Human Genome U133 Set 208109_PM_s_at - -
          Human Genome U133 Set 208109_s_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052423 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052423 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 174256 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1691501 NR_026813.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1691501 AF216224.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1691501 AL109679.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1691501 NR_026813.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1691501 AF216224.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1691501 AL109679.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1691501 NR_026813.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1691501 AF216224.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1691501 AL109679.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1691501 NR_026813.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1691501 AF216224.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1691501 AL109679.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1691501 NR_026813.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1691501 AF216224.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1691501 AL109679.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1691501 NR_026813.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1691501 AF216224.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1691501 AL109679.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1691501 NR_026813.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1691501 AF216224.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1691501 AL109679.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1691501 NR_026813.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1691501 AF216224.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1691501 AL109679.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline