Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00265057_m1
          See other IFNAR1 GE Assays ›
          Gene Symbol:
          IFNAR1
          Gene Name:
          interferon alpha and beta receptor subunit 1
          Gene Aliases:
          AVP, IFN-alpha-REC, IFNAR, IFNBR, IFRC
          Chromosome Location:
          Chr.21: 33324443 - 33360361 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00265057_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IFNAR1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000629.2 NP_000620.2 1 - 2 221 129
          XM_011529552.1 XP_011527854.1 1 - 2 221 129
          GenBank mRNA AK222770.1 - 1 - 2 157 129
          AK222812.1 - 1 - 2 157 129
          AK312631.1 - 1 - 2 157 129
          BC002590.2 - 1 - 2 136 129
          BC021825.1 - 1 - 2 153 4932084 129
          BG548299.1 - 1 - 2 226 129
          J03171.1 - 1 - 2 152 129
          JN543953.1 - 1 - 2 155 129
          JN543954.1 - 1 - 2 155 129
          JN543955.1 - 1 - 2 155 129
          JN543956.1 - 1 - 2 155 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100006960.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000128.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Immune Tolerance Network Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IFNAR1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          JAK-STAT cascade
          response to virus
          response to lipopolysaccharide
          positive regulation of interferon-beta production
          positive regulation of interferon-gamma production
          cellular response to interferon-alpha
          T cell activation
          type I interferon biosynthetic process
          positive regulation of transcription, DNA-templated
          positive regulation of interleukin-1 beta secretion
          regulation of peptidyl-tyrosine phosphorylation
          defense response to virus
          type I interferon signaling pathway
          regulation of type I interferon-mediated signaling pathway
          type I interferon receptor activity
          protein binding
          type I interferon binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR21000904.hg.1 ENST00000270139.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000904.hg.1 ENST00000442357.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000904.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000904.hg.1 TC21000128.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000493503.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 ENST00000493503.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000902.hg.1 ENST00000270139.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000902.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000902.hg.1 TC21000128.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21000905.hg.1 ENST00000270139.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000905.hg.1 ENST00000416947.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000905.hg.1 ENST00000442357.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000905.hg.1 ENST00000493503.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21000905.hg.1 NM_000629.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21000905.hg.1 TC21000128.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000493503.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 ENST00000493503.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay TC2100006960.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146738.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146738.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146738.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146738.hg.1 TC2100006960.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100146738.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146738.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146738.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146740.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146740.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146740.hg.1 ENST00000493503.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146740.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146740.hg.1 TC2100006960.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100146740.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146740.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146740.hg.1 ENST00000493503.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146740.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146736.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146736.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146736.hg.1 TC2100006960.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100146736.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146736.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146739.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146739.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146739.hg.1 NM_000629.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146739.hg.1 TC2100006960.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100146739.hg.1 ENST00000270139.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146739.hg.1 ENST00000442071.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146739.hg.1 NM_000629.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 225293 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0