Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00266002_m1
          See other PRMT1 GE Assays ›
          Gene Symbol:
          PRMT1
          Gene Name:
          protein arginine methyltransferase 1
          Gene Aliases:
          ANM1, HCP1, HRMT1L2, IR1B4
          Chromosome Location:
          Chr.19: 49676166 - 49688450 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00266002_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRMT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001536.5 NP_001527.3 2 - 3 223 59
          GenBank mRNA AL711069.1 - 2 - 3 117 59
          D66904.1 - 2 - 3 92 59
          Y10806.1 - 2 - 3 92 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900011781.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19002646.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PRMT1

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          protein methylation
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          cell surface receptor signaling pathway
          histone methylation
          peptidyl-arginine methylation
          peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
          neuron projection development
          histone H4-R3 methylation
          positive regulation of erythrocyte differentiation
          negative regulation of megakaryocyte differentiation
          positive regulation of hemoglobin biosynthetic process
          positive regulation of p38MAPK cascade
          protein binding
          methyltransferase activity
          N-methyltransferase activity
          protein methyltransferase activity
          methyl-CpG binding
          protein-arginine N-methyltransferase activity
          enzyme binding
          protein-arginine omega-N asymmetric methyltransferase activity
          histone methyltransferase activity
          identical protein binding
          histone methyltransferase activity (H4-R3 specific)
          poly(A) RNA binding
          mitogen-activated protein kinase p38 binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19010416.hg.1 TC19002646.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19010416.hg.1 ENST00000391851.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19010416.hg.1 ENST00000449059.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19010416.hg.1 ENST00000454376.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19010416.hg.1 ENST00000532489.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19010416.hg.1 NM_001207042.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19010416.hg.1 NM_001536.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19010416.hg.1 NM_198318.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19010416.hg.1 NR_033397.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000391851.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000454376.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000524771.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000525616.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000526224.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000527382.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000527412.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000528623.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000529284.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000529836.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000532489.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000534280.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000534465.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000534676.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000610806.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 NM_001207042.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 NM_001536.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 NM_198318.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 NR_033397.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 TC1900011781.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000391851.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000454376.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000524771.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000525616.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000526224.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000527382.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000527412.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000528623.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000529284.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000529836.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000532489.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000534280.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000534465.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000534676.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 ENST00000610806.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 NM_001207042.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 NM_001536.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 NM_198318.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900202086.hg.1 NR_033397.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 209837 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0