Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00269660_s1
          See other RHOB GE Assays ›
          Gene Symbol:
          RHOB
          Gene Name:
          ras homolog family member B
          Gene Aliases:
          ARH6, ARHB, MST081, MSTP081, RHOH6
          Chromosome Location:
          Chr.2: 20447071 - 20449445 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00269660_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RHOB
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004040.3 NP_004031.1 1 839 84
          GenBank mRNA AF498971.1 - 1 444 84
          AK124398.1 - 1 836 84
          AK312487.1 - 1 839 84
          BC066954.1 - 1 836 5264057 84
          BT019546.1 - 1 444 84
          BT019547.1 - 1 444 84
          CR542272.1 - 1 444 84
          M12174.1 - 1 361 84
          X06820.1 - 1 444 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200006891.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000116.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Human Phagocytosis of Microbes Human EGF Pathway HUMAN GPCR PATHWAY HUMAN GUIDANCE CUES AND GROWTH CONE MOTILITY HUMAN THROMBIN SIGNALING HUMAN RHO FAMILY GTPASES HUMAN RAS PATHWAY HUMAN PANCREATIC ADENOCARCINOMA 1700 Inventoried Human_EGF_Pathway Human_Phagocytosis_of_Microbes Human_Pancreatic_Adenocarcinoma Human_Guidance_Cues_and_Growth_Cone_Motility Human_GPCR_Pathway Human_Thrombin_Signaling Human_Ras_Pathway Human_Rho_Family_GTPases Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RHOB

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokinesis
          angiogenesis
          intracellular protein transport
          apoptotic process
          cell adhesion
          Rho protein signal transduction
          endosome to lysosome transport
          positive regulation of endothelial cell migration
          cell differentiation
          platelet activation
          regulation of cell migration
          negative regulation of cell migration
          positive regulation of apoptotic process
          positive regulation of angiogenesis
          negative regulation of cell cycle
          regulation of small GTPase mediated signal transduction
          endothelial tube morphogenesis
          cellular response to hydrogen peroxide
          cellular response to ionizing radiation
          GTPase activity
          protein binding
          GTP binding
          GDP binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02001317.hg.1 TC02000116.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02001317.hg.1 ENST00000272233.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02001317.hg.1 NM_004040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200006891.hg.1 ENST00000272233.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006891.hg.1 ENST00000272233.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006891.hg.1 ENST00000272233.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200006891.hg.1 NM_004040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200006891.hg.1 NM_004040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200006891.hg.1 NM_004040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02001316.hg.1 TC02000116.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02001316.hg.1 ENST00000272233.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02001316.hg.1 NM_004040.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02001318.hg.1 TC02000116.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02001318.hg.1 ENST00000272233.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02001318.hg.1 NM_004040.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200147032.hg.1 ENST00000272233.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200147032.hg.1 NM_004040.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200147032.hg.1 ENST00000272233.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200147032.hg.1 NM_004040.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200147032.hg.1 TC0200006891.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200147033.hg.1 ENST00000272233.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200147033.hg.1 NM_004040.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200147033.hg.1 ENST00000272233.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200147033.hg.1 NM_004040.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200147033.hg.1 TC0200006891.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200147034.hg.1 ENST00000272233.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200147034.hg.1 NM_004040.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200147034.hg.1 ENST00000272233.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200147034.hg.1 NM_004040.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200147034.hg.1 TC0200006891.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0200006891.hg.1 ENST00000272233.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200006891.hg.1 ENST00000272233.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200006891.hg.1 ENST00000272233.5 Ensembl
          Affymetrix Human Clariom D Assay TC0200006891.hg.1 NM_004040.3 Refseq
          Affymetrix Human Clariom D Assay TC0200006891.hg.1 NM_004040.3 Refseq
          Affymetrix Human Clariom D Assay TC0200006891.hg.1 NM_004040.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 149591 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0