Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00287613_m1
          See other GALNT13 GE Assays ›
          Gene Symbol:
          GALNT13
          Gene Name:
          polypeptide N-acetylgalactosaminyltransferase 13
          Gene Aliases:
          GalNAc-T13
          Chromosome Location:
          Chr.2: 153871898 - 154454177 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00287613_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GALNT13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001301627.1 NP_001288556.1 4 - 5 790 62
          NM_052917.3 NP_443149.2 6 - 7 1268 62
          XM_011510537.2 XP_011508839.1 6 - 7 1263 62
          XM_011510538.2 XP_011508840.1 4 - 5 675 62
          XM_017003258.1 XP_016858747.1 6 - 7 1117 62
          XM_017003259.1 XP_016858748.1 5 - 6 842 62
          XM_017003260.1 XP_016858749.1 6 - 7 1263 62
          XM_017003261.1 XP_016858750.1 6 - 7 1127 62
          XM_017003262.1 XP_016858751.1 4 - 5 533 62
          XM_017003263.1 XP_016858752.1 4 - 5 675 62
          GenBank mRNA AB067505.1 - 4 - 5 568 62
          AB078142.1 - 4 - 5 686 62
          AJ505991.2 - 4 - 5 686 62
          AK131195.1 - 6 - 7 1213 62
          AK308675.1 - 4 - 5 675 62
          BC101031.1 - 4 - 5 686 40007710 62
          BC101032.2 - 4 - 5 686 40007711 62
          BC101033.1 - 4 - 5 686 40007712 62
          BC101034.2 - 4 - 5 686 40007713 62
          KJ534843.1 - 4 - 5 686 62
          KJ535058.1 - 3 - 4 517 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200009700.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000944.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GALNT13

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          O-glycan processing
          polypeptide N-acetylgalactosaminyltransferase activity
          carbohydrate binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000392825.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000409237.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000422126.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000422126.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000422126.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000422126.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000422126.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000422126.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000431076.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000431076.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000431076.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000431076.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000431076.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000431076.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000431076.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000431076.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000431076.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000431076.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000431076.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000434213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000434213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000434213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000434213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000434213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000450838.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000450838.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000450838.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000450838.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000450838.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000450838.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000453715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000453715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000453715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000453715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000453715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000453715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000453715.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000487047.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000487047.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000487047.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000487047.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000487047.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000487047.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000487047.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000489553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000489553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000489553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000489553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000489553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000494805.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000494805.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000494805.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000494805.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 ENST00000494805.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_001301627.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009700.hg.1 NM_052917.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline