Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00293736_m1
          See other INKA1 GE Assays ›
          Gene Symbol:
          INKA1
          Gene Name:
          inka box actin regulator 1
          Gene Aliases:
          C3orf54, FAM212A, INCA
          Chromosome Location:
          Chr.3: 49803261 - 49805030 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00293736_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: INKA1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_203370.2 NP_976248.2 1 - 2 183 109
          GenBank mRNA BC012170.1 - 1 - 2 190 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300007392.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000299.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene INKA1

          Gene Ontology Categories:

          Process(es)

          protein binding
          protein kinase binding
          protein serine/threonine kinase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02228294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02228303 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15436463 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02228299 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02228308 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02228307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02228300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P22362900 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 419408 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3239267 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P158470 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 12382 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 21522 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P30997713 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3239267 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3239267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228306 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228307 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228301 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15436463 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228303 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22362905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228297 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228298 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228299 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22362900 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228299 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228307 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228306 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22362905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228294 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228297 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15436463 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228301 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228298 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22362900 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02228303 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P158470 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02228307 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02228306 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02228299 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02228298 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02228302 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P02228297 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15436460 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15436459 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15436461 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02228297 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02228299 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02228298 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15436458 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P158470 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228370 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228283 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228398 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228343 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228296 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228372 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228299 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228385 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228374 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228392 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228241 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228255 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228380 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228263 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228287 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228345 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228239 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228367 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228246 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228298 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228281 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228302 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228292 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15436455 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228242 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228351 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228257 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15436463 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228341 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15436458 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228360 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228377 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228269 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228306 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228273 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228277 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228307 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228252 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228347 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228354 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228390 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228301 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228291 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228362 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228245 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228261 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228285 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228375 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228297 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228271 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228396 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228356 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228264 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228267 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228387 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02228258 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_203370_at - -
          Affymetrix Human Gene 1.0 ST Array 8079840 - -
          Affymetrix Human Gene 1.0 ST Array 8079841 - -
          Affymetrix Human Gene 1.1 ST Array 8079839 - -
          Affymetrix HT HG-U133+ PM Array Plate 229507_PM_at - -
          Affymetrix Human Genome U219 Array 11724713_at - -
          Affymetrix Human Genome U219 Array 11724713_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR03006191.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03006191.hg.1 NM_203370.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03006191.hg.1 TC03000299.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03006190.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03006190.hg.1 NM_203370.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03006190.hg.1 TC03000299.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0300007392.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007392.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007392.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007392.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300007392.hg.1 NM_203370.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007392.hg.1 NM_203370.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007392.hg.1 NM_203370.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300007392.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300152604.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300152604.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300152604.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300152604.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300152604.hg.1 TC0300007392.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300152605.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300152605.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300152605.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300152605.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300152605.hg.1 TC0300007392.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0300007392.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007392.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007392.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007392.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300007392.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay TC0300007392.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay TC0300007392.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay TC0300007392.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300152606.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300152606.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300152606.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300152606.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300152606.hg.1 TC0300007392.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300152607.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300152607.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300152607.hg.1 ENST00000333323.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300152607.hg.1 NM_203370.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300152607.hg.1 TC0300007392.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 229507_at - -
          Human Genome U133 Plus 2.0 Array 229507_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_203370_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229507_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229507_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229507_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229507_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229507_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229507_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229507_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229507_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229507_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229507_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 229507_at4 - -
          Human Genome U95 Set 47115_PM_at - -
          Human Genome U95 Set 47115_at - -
          Human Genome U133 Set 229507_at - -
          Human Genome U133 Set 229507_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A022628 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A022628 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 108077 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1690454 NM_203370.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1690454 BC012170.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1690454 NM_203370.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1690454 BC012170.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1690454 NM_203370.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1690454 BC012170.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1690454 NM_203370.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1690454 BC012170.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1690454 NM_203370.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1690454 BC012170.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_42766423-S NM_203370.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_42766423-S BC012170.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_28059-S - -
          Illumina human-6 v1.0 expression beadchip 0000940397 - -
          Illumina humanRef-8 v1.0 expression beadchip 0000940397 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1690454 NM_203370.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1690454 BC012170.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1690454 NM_203370.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1690454 BC012170.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 2710678 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_28059 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1690454 NM_203370.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1690454 BC012170.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1690454 NM_203370.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1690454 BC012170.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1690454 NM_203370.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1690454 BC012170.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1690454 NM_203370.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1690454 BC012170.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1690454 NM_203370.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1690454 BC012170.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline