Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00366272_m1
          See other TNFRSF10B GE Assays ›
          Gene Symbol:
          TNFRSF10B
          Gene Name:
          TNF receptor superfamily member 10b
          Gene Aliases:
          CD262, DR5, KILLER, KILLER/DR5, TRAIL-R2, TRAILR2, TRICK2, TRICK2A, TRICK2B, TRICKB, ZTNFR9
          Chromosome Location:
          Chr.8: 23020133 - 23069031 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00366272_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TNFRSF10B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003842.5 NP_003833.4 2 - 3 392 103
          NM_147187.3 NP_671716.2 2 - 3 392 103
          GenBank mRNA AB451272.1 - 2 - 3 255 103
          AB451399.1 - 2 - 3 255 103
          AF012535.2 - 2 - 3 394 103
          AF012628.1 - 2 - 3 255 103
          AF016266.1 - 2 - 3 372 103
          AF016268.1 - 2 - 3 364 103
          AF016849.1 - 2 - 3 399 103
          AF018657.1 - 2 - 3 255 103
          AF018658.1 - 2 - 3 255 103
          AF020501.1 - 2 - 3 255 103
          AF022386.1 - 2 - 3 255 103
          AF153687.1 - 2 - 3 373 103
          AF192548.1 - 2 - 3 355 103
          AK291216.1 - 2 - 3 375 103
          AL529460.3 - 2 - 3 548 103
          AY358277.1 - 2 - 3 394 103
          BC001281.1 - 2 - 3 402 103
          BX538104.1 - 2 - 3 388 103
          CR541898.1 - 2 - 3 255 103
          DA738472.1 - 2 - 3 548 103
          DA889488.1 - 2 - 3 392 103

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Human NFkB Pathway Human Apoptosis Inventoried Human_Nfkb Human_Apoptosis Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TNFRSF10B

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response to tumor cell
          apoptotic process
          signal transduction
          cell surface receptor signaling pathway
          activation of NF-kappaB-inducing kinase activity
          extrinsic apoptotic signaling pathway via death domain receptors
          response to endoplasmic reticulum stress
          TRAIL-activated apoptotic signaling pathway
          regulation of apoptotic process
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to mechanical stimulus
          death receptor activity
          protein binding
          TRAIL receptor activity
          signaling receptor activity
          TRAIL binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0