Search
Search

HIST1H2AG| Gene Symbol: HIST1H2AG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021064.4 | NP_066408.1 | 1 | 454 | 76 | |
| GenBank mRNA | BC016677.1 | - | 1 | 439 | 4342191 | 76 |
| L19778.1 | - | 1 | 450 | 76 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817564 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627578 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817569 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627572 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627584 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627576 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627577 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627583 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24817568 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627574 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627579 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04627582 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P414658 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P414658 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P303354 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 36698 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 40956 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P122277 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P134794 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P122277 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P134794 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 7186 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3362320 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817568 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627582 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627574 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627584 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627572 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627572 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627582 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627574 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04627584 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24817568 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P303354 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P122277 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P134794 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627581 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627587 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627580 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627577 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04627575 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P122277 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P134794 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P134794 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P414658 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P303354 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 5081 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 1073 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 8122 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 22080 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 29548 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P122277 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627616 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627594 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627577 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627583 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627581 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627574 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627580 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04627585 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_021064_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8117536 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8117535 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 207156_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11759131_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11734234_at | - | - |
| Affymetrix Human Genome U219 Array | 11734234_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11759131_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014083.hg.1 | ENST00000359193.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014083.hg.1 | ENST00000359193.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014083.hg.1 | ENST00000359193.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014083.hg.1 | NM_021064.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014083.hg.1 | NM_021064.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600197768.hg.1 | ENST00000359193.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600197768.hg.1 | NM_021064.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600197768.hg.1 | ENST00000359193.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600197768.hg.1 | NM_021064.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600197768.hg.1 | TC0600014083.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014083.hg.1 | ENST00000359193.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014083.hg.1 | ENST00000359193.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014083.hg.1 | ENST00000359193.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014083.hg.1 | NM_021064.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014083.hg.1 | NM_021064.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600197769.hg.1 | ENST00000359193.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600197769.hg.1 | ENST00000359193.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600197769.hg.1 | TC0600014083.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 214554_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208583_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214554_at | - | - |
| Human Genome U133 Plus 2.0 Array | 207156_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 207156_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208583_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214542_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214542_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 214554_PM_at | - | - |
| Human Genome U133A 2.0 Array | 208583_x_at | - | - |
| Human Genome U133A 2.0 Array | 214542_x_at | - | - |
| Human Genome U133A 2.0 Array | 207156_PM_at | - | - |
| Human Genome U133A 2.0 Array | 207156_at | - | - |
| Human Genome U133A 2.0 Array | 208583_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 214542_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 214554_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_021064_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw598955 | - | - |
| Human Genome U95 Set | 284_at | - | - |
| Human Genome U95 Set | 284_PM_at | - | - |
| Human Genome U95 Set | 34157_f_at | - | - |
| Human Genome U95 Set | 34156_PM_i_at | - | - |
| Human Genome U95 Set | 34157_PM_f_at | - | - |
| Human Genome U95 Set | 31768_PM_at | - | - |
| Human Genome U95 Set | 285_g_at | - | - |
| Human Genome U95 Set | 31768_at | - | - |
| Human Genome U95 Set | 285_PM_g_at | - | - |
| Human Genome U95 Set | 34156_i_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207156_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207156_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207156_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207156_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207156_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207156_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207156_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207156_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207156_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207156_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 207156_at7 | - | - |
| Human Genome U133 Set | 214554_PM_at | - | - |
| Human Genome U133 Set | 207156_at | - | - |
| Human Genome U133 Set | 207156_PM_at | - | - |
| Human Genome U133 Set | 208583_PM_x_at | - | - |
| Human Genome U133 Set | 214554_at | - | - |
| Human Genome U133 Set | 214542_x_at | - | - |
| Human Genome U133 Set | 214542_PM_x_at | - | - |
| Human Genome U133 Set | 208583_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031138 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031138 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 224381 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1686478 | BC016677.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1686478 | BC067782.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1686478 | L19778.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2184602 | BC016677.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2184602 | BC067782.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2184602 | L19778.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1686478 | BC016677.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1686478 | BC067782.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1686478 | L19778.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2184602 | BC016677.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2184602 | BC067782.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2184602 | L19778.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1686478 | BC016677.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1686478 | BC067782.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1686478 | L19778.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1686478 | BC016677.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1686478 | BC067782.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1686478 | L19778.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2184602 | BC016677.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2184602 | BC067782.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2184602 | L19778.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1686478 | BC016677.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1686478 | BC067782.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1686478 | L19778.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2184602 | BC016677.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2184602 | BC067782.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2184602 | L19778.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_38570130-S | BC016677.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_38570130-S | BC067782.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_38570130-S | L19778.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_38570130-S | NM_021064.4 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_13095-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0005080484 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0005080484 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1686478 | BC016677.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1686478 | BC067782.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1686478 | L19778.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1686478 | BC016677.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1686478 | BC067782.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1686478 | L19778.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 1940064 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_13095 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2184602 | BC016677.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2184602 | BC067782.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2184602 | L19778.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1686478 | BC016677.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1686478 | BC067782.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1686478 | L19778.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2184602 | BC016677.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2184602 | BC067782.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2184602 | L19778.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2184602 | BC016677.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2184602 | BC067782.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2184602 | L19778.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1686478 | BC016677.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1686478 | BC067782.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1686478 | L19778.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1686478 | BC016677.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1686478 | BC067782.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1686478 | L19778.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2184602 | BC016677.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2184602 | BC067782.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2184602 | L19778.1 | human_transcriptome |