Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00394952_m1
          See other SCN2B GE Assays ›
          Gene Symbol:
          SCN2B
          Gene Name:
          sodium voltage-gated channel beta subunit 2
          Gene Aliases:
          ATFB14
          Chromosome Location:
          Chr.11: 118162806 - 118176639 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00394952_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SCN2B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004588.5 NP_004579.1 3 - 4 656 86
          GenBank mRNA AF007783.1 - 3 - 4 448 86
          AF049498.1 - 3 - 4 620 86
          AF107028.1 - 3 - 4 659 86
          AK313891.1 - 3 - 4 625 86
          AY358945.1 - 3 - 4 593 86
          BC036793.1 - 3 - 4 616 86
          BT019487.1 - 3 - 4 448 86
          U87555.1 - 3 - 4 561 86

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Human Cholera Infection Inventoried Human_Cholera_Infection Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SCN2B

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transport
          sodium ion transport
          chemical synaptic transmission
          nervous system development
          response to heat
          gene expression
          positive regulation of sodium ion transport
          sodium ion transmembrane transport
          response to pyrethroid
          cardiac muscle contraction
          regulation of atrial cardiac muscle cell membrane depolarization
          cardiac conduction
          cardiac muscle cell action potential involved in contraction
          membrane depolarization during action potential
          membrane depolarization during cardiac muscle cell action potential
          regulation of heart rate by cardiac conduction
          membrane repolarization during ventricular cardiac muscle cell action potential
          voltage-gated sodium channel activity
          sodium channel regulator activity
          voltage-gated sodium channel activity involved in cardiac muscle cell action potential
          voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 ENST00000278947.5 Ensembl
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 NM_004588.4 Refseq
          Affymetrix Human Clariom D Assay TC1100012470.hg.1 NM_004588.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline