Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00400616_m1
          See other CRYBG3 GE Assays ›
          Gene Symbol:
          CRYBG3
          Gene Name:
          crystallin beta-gamma domain containing 3
          Gene Aliases:
          DKFZp667G2110, vlAKAP
          Chromosome Location:
          Chr.3: 97822011 - 97944984 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00400616_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CRYBG3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153605.4 NP_705833.3 16 - 17 8309 102
          XM_005247117.5 XP_005247174.1 15 - 16 8242 102
          GenBank mRNA AF451988.1 - 4 - 5 409 102
          AK091199.1 - 8 - 9 751 102
          AL831927.1 - 4 - 5 303 102
          BC050394.1 - 1 - 2 63 102
          BX647079.1 - 8 - 9 768 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300008088.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CRYBG3

          Gene Ontology Categories:

          Function(s) Process(es)

          lens development in camera-type eye
          visual perception
          structural constituent of eye lens
          carbohydrate binding
          protein kinase A binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 141118 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 94778 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 168006 AK092236.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 168006 AK094139.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 168006 AK122749.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 168006 AK122766.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 168006 BC060863.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 168006 NM_153263.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CGH Microarray 4x180K 49623 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 47248 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 3101 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 816173 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 541136 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 858822 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 181226 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 50631 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 404852 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 265404 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 343862 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 211225 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 485386 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 831079 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 594797 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 851220 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 615955 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 791833 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 66365 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 739518 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 26805 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 888494 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 474401 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 372210 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P303810 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P303810 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P303815 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P920851 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00766201 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P36371460 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P36371436 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00766195 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P36371453 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P118560 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P36371376 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P36371487 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P16328298 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00766130 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8081186 - -
          Affymetrix Human Gene 1.0 ST Array 8081191 - -
          Affymetrix Human Gene 1.0 ST Array 8081180 - -
          Affymetrix Human Gene 1.0 ST Array 8081187 - -
          Affymetrix Human Gene 1.0 ST Array 8081185 - -
          Affymetrix Human Gene 1.0 ST Array 8081184 - -
          Affymetrix Human Gene 1.0 ST Array 8081189 - -
          Affymetrix Human Gene 1.0 ST Array 8081181 - -
          Affymetrix Human Gene 1.0 ST Array 8081182 - -
          Affymetrix Human Gene 1.0 ST Array 8081190 - -
          Affymetrix Human Gene 1.0 ST Array 8081188 - -
          Affymetrix Human Gene 1.0 ST Array 8081179 - -
          Affymetrix Human Gene 1.0 ST Array 8081177 - -
          Affymetrix Human Gene 1.0 ST Array 8081183 - -
          Affymetrix Human Gene 1.0 ST Array 8081178 - -
          Affymetrix Human Gene 1.1 ST Array 8081171 - -
          Affymetrix HT HG-U133+ PM Array Plate 214030_PM_at - -
          Affymetrix Human Clariom D Assay PSR0300157687.hg.1 ENST00000389622.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300157687.hg.1 NM_153605.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300157687.hg.1 ENST00000389622.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300157687.hg.1 NM_153605.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300157687.hg.1 TC0300008088.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 214030_at - -
          Human Genome U133 Plus 2.0 Array 214030_PM_at - -
          Human Genome U133A 2.0 Array 214030_at - -
          Human Genome U133A 2.0 Array 214030_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw511234 - -
          Human Genome U95 Set 41274_at - -
          Human Genome U95 Set 41274_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214030_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214030_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214030_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214030_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214030_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214030_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214030_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214030_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214030_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214030_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214030_at8 - -
          Human Genome U133 Set 214030_PM_at - -
          Human Genome U133 Set 214030_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 105842 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2190684 CR749313.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2190684 BX647079.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2190684 BX648353.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2190684 BC036247.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2190684 AK098569.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2190684 CR749313.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2190684 BX647079.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2190684 BX648353.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2190684 BC036247.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2190684 AK098569.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2190684 CR749313.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2190684 BX647079.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2190684 BX648353.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2190684 BC036247.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2190684 AK098569.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2190684 CR749313.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2190684 BX647079.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2190684 BX648353.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2190684 BC036247.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2190684 AK098569.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2190684 CR749313.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2190684 BX647079.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2190684 BX648353.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2190684 BC036247.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2190684 AK098569.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2190684 CR749313.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2190684 BX647079.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2190684 BX648353.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2190684 BC036247.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2190684 AK098569.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline