Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00416846_m1
          See other IFITM5 GE Assays ›
          Gene Symbol:
          IFITM5
          Gene Name:
          interferon induced transmembrane protein 5
          Gene Aliases:
          BRIL, DSPA1, Hrmp1, OI5, fragilis4
          Chromosome Location:
          Chr.11: 298200 - 299526 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00416846_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IFITM5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025295.2 NP_001020466.1 1 - 2 223 58
          GenBank mRNA BC150562.1 - 1 - 2 216 40127610 58
          BC150563.1 - 1 - 2 216 40127611 58
          CR747200.1 - 1 - 2 225 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100013119.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11003502.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IFITM5

          Gene Ontology Categories:

          Function(s)

          in utero embryonic development
          response to biotic stimulus
          bone mineralization
          regulation of bone mineralization
          bone morphogenesis

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594339 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27988152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594336 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594338 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594342 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594351 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594345 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594353 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27988155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594343 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594355 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594352 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594349 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594347 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27988148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27988145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594354 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27988147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594335 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27988164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07594346 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27988151 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3352995 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P150664 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 43673 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28585 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02378583 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P02378581 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3352995 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3352995 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594355 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594343 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594352 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594338 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988153 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594342 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594349 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988153 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594349 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988151 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594343 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594352 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594338 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594355 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27988164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07594342 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P150664 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07594357 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07594364 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07594354 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07594358 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07594356 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07594360 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07594363 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P07594349 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P07594357 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16405581 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16405589 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P07594350 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16405582 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16405590 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P16405591 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P07594351 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P150664 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02378583 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594340 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405539 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594304 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594316 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594341 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405545 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594364 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594343 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594301 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594360 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594357 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405532 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405546 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405574 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405579 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594306 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594355 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594336 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594303 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405519 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594311 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405522 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594314 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405570 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405590 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405517 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594338 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405525 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594354 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594308 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405583 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594344 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594349 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594307 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405584 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594315 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594358 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405541 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594342 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594313 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405582 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594320 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405533 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16405531 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594347 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594346 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07594363 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001025295_at - -
          Affymetrix Human Gene 1.0 ST Array 7945370 - -
          Affymetrix Human Gene 1.0 ST Array 7945369 - -
          Affymetrix Human Gene 1.1 ST Array 7945368 - -
          Affymetrix Human Genome U219 Array 11737383_PM_at - -
          Affymetrix Human Genome U219 Array 11737383_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR11015374.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11015374.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11015374.hg.1 TC11003502.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11015375.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11015375.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11015375.hg.1 TC11003502.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100013119.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013119.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Clariom D Assay TC1100013119.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100203127.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100203127.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100203127.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100203127.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100203127.hg.1 TC1100013119.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100203126.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100203126.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100203126.hg.1 ENST00000382614.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100203126.hg.1 NM_001025295.2 Refseq
          Affymetrix Human Clariom D Assay PSR1100203126.hg.1 TC1100013119.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A044710 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A044710 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 163015 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1711032 NM_001025295.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1711032 BC150562.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1711032 BC150563.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1720992 - -
          HumanHT-12 V4.0 expression beadchip ILMN_1711032 NM_001025295.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1711032 BC150562.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1711032 BC150563.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1711032 NM_001025295.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1711032 BC150562.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1711032 BC150563.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1711032 NM_001025295.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1711032 BC150562.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1711032 BC150563.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_27022-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1720992 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1720992 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 5560162 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_27022 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1711032 NM_001025295.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1711032 BC150562.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1711032 BC150563.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1711032 NM_001025295.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1711032 BC150562.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1711032 BC150563.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1711032 NM_001025295.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1711032 BC150562.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1711032 BC150563.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1720992 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1711032 NM_001025295.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1711032 BC150562.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1711032 BC150563.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline