Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00420249_m1
          See other LINC01549 GE Assays ›
          Gene Symbol:
          LINC01549
          Gene Name:
          long intergenic non-protein coding RNA 1549
          Gene Aliases:
          C21orf37
          Chromosome Location:
          Chr.21: 17438890 - 17449185 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00420249_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01549
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BX089409.1 - 1 - 2 112 90
          N47348.1 - 1 - 2 113 90

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17222193 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944184 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11285985 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944174 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944214 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944199 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944165 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944185 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944191 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944144 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286057 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286070 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11285989 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944180 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944177 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944221 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286003 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286005 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286046 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944193 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944202 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11285990 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286073 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11285987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286014 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11285986 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286011 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17222188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11286045 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31944164 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3781094 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P15863 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 7203 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 30791 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3781094 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P15863 - -
          Array Name Probe IDs Targets Target Type
          Human Genome U133 Plus 2.0 Array 240801_at - -
          Human Genome U133 Plus 2.0 Array 240801_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240801_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240801_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240801_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240801_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240801_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240801_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240801_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240801_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240801_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240801_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240801_at11 - -
          Human Genome U95 Set 45989_at - -
          Human Genome U95 Set 45989_PM_at - -
          Human Genome U133 Set 240801_at - -
          Human Genome U133 Set 240801_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019072 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019072 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 136025 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline