Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00541779_s1
          See other FLJ30679 GE Assays ›
          Gene Symbol:
          FLJ30679
          Gene Name:
          uncharacterized protein FLJ30679
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 86555320 - 86557299 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00541779_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLJ30679
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026674.1 1 1246 55
          GenBank mRNA AK055241.1 - 1 1248 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600008737.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000669.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 100327 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 40376 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 288267 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P346327 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P346327 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 7791 - -
          Agilent Human Genome CGH Microarray 44B A_14_P124164 - -
          Agilent Human Genome CGH Microarray 44A A_14_P124164 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 61862 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P346327 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P346327 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16923121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30713375 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16923123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16923122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10170617 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10170613 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10170612 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30713377 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16923121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30713377 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16923123 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10170613 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16923122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10170612 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30713375 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10170617 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P346327 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P124164 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P124164 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P124164 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P346327 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 4503 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 45194 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P124164 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170637 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170640 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170644 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170618 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170613 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170632 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170642 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170662 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170635 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170655 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170656 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170638 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170650 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170629 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170659 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170619 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170615 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16923140 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170649 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16923120 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16923121 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170664 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170646 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10170652 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P124164 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7997732 - -
          Affymetrix Human Gene 1.1 ST Array 7997731 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553258_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1600008737.hg.1 ENST00000624773.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008737.hg.1 NR_026674.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16009456.hg.1 NR_026674.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16009456.hg.1 TC16000669.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1600165243.hg.1 ENST00000624773.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165243.hg.1 NR_026674.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165243.hg.1 ENST00000624773.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165243.hg.1 NR_026674.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165243.hg.1 TC1600008737.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1600008737.hg.1 ENST00000624773.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008737.hg.1 NR_026674.1 Refseq
          Human Genome U133 Plus 2.0 Array 1553258_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553258_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553258_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553258_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553258_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553258_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553258_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553258_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553258_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553258_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553258_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553258_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553258_at6 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054876 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054876 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 221551 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1711772 AK055241.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1711772 NR_026674.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1711772 AK055241.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1711772 NR_026674.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1711772 AK055241.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1711772 NR_026674.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1711772 AK055241.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1711772 NR_026674.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1711772 AK055241.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1711772 NR_026674.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline