Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00544762_m1
          See other PSMB9 GE Assays ›
          Gene Symbol:
          PSMB9
          Gene Name:
          proteasome subunit beta 9
          Gene Aliases:
          LMP2, PSMB6i, RING12, beta1i
          Chromosome Location:
          Chr.6: 32854161 - 32859851 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00544762_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PSMB9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002800.4 NP_002791.1 2 - 3 201 102
          GenBank mRNA BC065513.1 - 2 - 3 156 5749985 102
          BI832878.1 - 2 - 3 183 102
          BM998687.1 - 4 - 5 102
          CR541656.1 - 2 - 3 132 102
          S75169.1 - 1 - 2 111 102
          U01025.1 - 1 - 2 111 102
          X62741.1 - 2 - 3 147 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600014110.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06004082.hg.1, TC6_apd_hap1000122.hg.1, TC6_cox_hap2000259.hg.1, TC6_dbb_hap3000226.hg.1, TC6_mann_hap4000190.hg.1, TC6_mcf_hap5000202.hg.1, TC6_qbl_hap6000215.hg.1, TC6_ssto_hap7000193.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Human Antigen Processing and Presantation by MHCS HUMAN DNA REPAIR MECHANISM HUMAN SMAD SIGNALING NETWORK HUMAN HUNTINGTON DISEASE PATHWAY Inventoried Human_Antigen_Processing_and_Presentation_by_MHCS Human_Huntington_Disease_Pathway Human_DNA_Repair_Mechanism Human_SMAD_Signaling_Network Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PSMB9

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein polyubiquitination
          liver development
          stimulatory C-type lectin receptor signaling pathway
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          regulation of cellular amino acid metabolic process
          muscle atrophy
          anaphase-promoting complex-dependent catabolic process
          tumor necrosis factor-mediated signaling pathway
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          response to drug
          proteasome-mediated ubiquitin-dependent protein catabolic process
          response to alkaloid
          regulation of mRNA stability
          spleen development
          thymus development
          T cell receptor signaling pathway
          negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          Wnt signaling pathway, planar cell polarity pathway
          cellular response to electrical stimulus
          cellular response to interleukin-1
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          cellular response to virus
          response to benzene
          regulation of cysteine-type endopeptidase activity
          threonine-type endopeptidase activity
          protein binding
          proteasome binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1000681.hg.1 TC6_apd_hap1000122.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1000681.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3001805.hg.1 TC6_dbb_hap3000226.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3001805.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6001757.hg.1 TC6_qbl_hap6000215.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6001757.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2001794.hg.1 TC6_cox_hap2000259.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2001794.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5001582.hg.1 TC6_mcf_hap5000202.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5001582.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7001615.hg.1 TC6_ssto_hap7000193.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7001615.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4001403.hg.1 TC6_mann_hap4000190.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4001403.hg.1 NM_002800.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06004709.hg.1 TC06004082.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06004709.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06004709.hg.1 NM_002800.4 Refseq
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Clariom D Assay TC0600014110.hg.1 NM_002800.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600198419.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198419.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198419.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198419.hg.1 NM_002800.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600198419.hg.1 ENST00000374859.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198419.hg.1 ENST00000464863.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198419.hg.1 ENST00000467593.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600198419.hg.1 NM_002800.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600198419.hg.1 TC0600014110.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0