Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00600918_s1
          See other MRGPRX1 GE Assays ›
          Gene Symbol:
          MRGPRX1
          Gene Name:
          MAS related GPR family member X1
          Gene Aliases:
          GPCR, MGRG2, MRGX1, SNSR4
          Chromosome Location:
          Chr.11: 18933813 - 18935002 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00600918_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MRGPRX1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147199.3 NP_671732.3 1 454 93
          GenBank mRNA BC096835.1 - 1 462 30706612 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100010284.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001474.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MRGPRX1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          acute-phase response
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28074060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16425518 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07673860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28074021 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16425526 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16425538 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07673859 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07673853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16425517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07673864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07673855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07673851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16425579 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28074019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16425559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07673849 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07673850 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28074037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28074080 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07673870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28074065 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07673847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07673861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28074079 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 422679 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 312027 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 366962 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P365725 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P365725 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 26403 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 42687 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P32867432 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P365725 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P365725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07673870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16425565 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16425559 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16425573 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16425594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07673874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074071 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074079 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07673867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07673872 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16425579 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07673866 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16425573 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07673867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07673870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07673874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16425579 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074079 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16425559 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07673872 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16425594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074071 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28074069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07673866 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16425565 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P365725 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07673868 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07673876 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07673870 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07673866 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07673867 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07673865 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07673874 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07673875 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07673872 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07673869 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07673873 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P365725 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02403838 - -
          Agilent Human 1B Microarray G4111A A_32_P106919 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07673847 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16425559 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07673855 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07673850 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07673870 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07673866 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07673864 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07673869 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07673852 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07673867 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07673872 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02403838 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_147199_at - -
          Affymetrix Human Gene 1.0 ST Array 7947094 - -
          Affymetrix Human Gene 1.0 ST Array 7947095 - -
          Affymetrix Human Gene 1.1 ST Array 7947093 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553401_PM_at - -
          Affymetrix Human Genome U219 Array 11735426_at - -
          Affymetrix Human Genome U219 Array 11735426_PM_at - -
          Affymetrix Human Genome U219 Array 11735427_x_at - -
          Affymetrix Human Genome U219 Array 11735427_PM_x_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1100010284.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010284.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010284.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010284.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010284.hg.1 ENST00000526914.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010284.hg.1 ENST00000526914.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010284.hg.1 ENST00000526914.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100010284.hg.1 NM_147199.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010284.hg.1 NM_147199.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010284.hg.1 NM_147199.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100010284.hg.1 NM_147199.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11018679.hg.1 ENST00000302797.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11018679.hg.1 NM_147199.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11018679.hg.1 TC11001474.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100178275.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100178275.hg.1 ENST00000526914.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100178275.hg.1 NM_147199.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100178275.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100178275.hg.1 ENST00000526914.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100178275.hg.1 NM_147199.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100178275.hg.1 TC1100010284.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100178277.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100178277.hg.1 NM_147199.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100178277.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100178277.hg.1 NM_147199.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100178277.hg.1 TC1100010284.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100010284.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100010284.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100010284.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100010284.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100010284.hg.1 ENST00000526914.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010284.hg.1 ENST00000526914.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010284.hg.1 ENST00000526914.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100010284.hg.1 NM_147199.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010284.hg.1 NM_147199.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010284.hg.1 NM_147199.3 Refseq
          Affymetrix Human Clariom D Assay TC1100010284.hg.1 NM_147199.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100178276.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100178276.hg.1 ENST00000526914.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100178276.hg.1 NM_147199.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100178276.hg.1 ENST00000302797.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100178276.hg.1 ENST00000526914.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100178276.hg.1 NM_147199.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100178276.hg.1 TC1100010284.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1553401_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553401_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_147199_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553401_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553401_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553401_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553401_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553401_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553401_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553401_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553401_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553401_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553401_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553401_at7 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045003 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045003 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 227948 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1773156 NM_147199.3 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1773156 BC096835.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1773156 NM_147199.3 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1773156 BC096835.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1773156 NM_147199.3 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1773156 BC096835.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1773156 NM_147199.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1773156 BC096835.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1773156 NM_147199.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1773156 BC096835.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_22218356-S NM_147199.3 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_22218356-S BC096835.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_22257-S - -
          Illumina human-6 v1.0 expression beadchip 0001340047 - -
          Illumina humanRef-8 v1.0 expression beadchip 0001340047 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1773156 NM_147199.3 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1773156 BC096835.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1773156 NM_147199.3 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1773156 BC096835.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 5890603 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_22257 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1773156 NM_147199.3 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1773156 BC096835.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1773156 NM_147199.3 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1773156 BC096835.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1773156 NM_147199.3 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1773156 BC096835.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1773156 NM_147199.3 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1773156 BC096835.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1773156 NM_147199.3 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1773156 BC096835.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline