Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00603550_g1
          See other TUBB2B GE Assays ›
          Gene Symbol:
          TUBB2B
          Gene Name:
          tubulin beta 2B class IIb
          Gene Aliases:
          PMGYSA, bA506K6.1
          Chromosome Location:
          Chr.6: 3224261 - 3227734 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00603550_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TUBB2B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178012.4 NP_821080.1 2 - 3 358 130
          GenBank mRNA AK289433.1 - 2 - 3 279 130
          BC001352.2 - 2 - 3 358 2964590 130
          BC063610.1 - 2 - 3 261 4941267 130
          BT019930.1 - 2 - 3 167 130
          CR456788.1 - 2 - 3 167 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600010572.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001228.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TUBB2B

          Panther Classification:

          Molecular Function -

          tubulin

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          cytoskeleton organization
          microtubule-based process
          GTPase activity
          structural constituent of cytoskeleton
          protein binding
          GTP binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600010567.hg.1 ENST00000404155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010567.hg.1 ENST00000404155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010567.hg.1 ENST00000404155.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014584.hg.1 TC06001228.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06014584.hg.1 ENST00000259818.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014584.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014581.hg.1 TC06001228.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06014581.hg.1 ENST00000259818.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014581.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014581.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014582.hg.1 TC06001228.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06014582.hg.1 ENST00000259818.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014582.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014582.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014580.hg.1 TC06001228.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06014580.hg.1 ENST00000259818.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014580.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014580.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06014583.hg.1 TC06001228.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06014583.hg.1 ENST00000259818.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06014583.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171835.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171835.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171835.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171835.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171835.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171835.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171835.hg.1 TC0600010572.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay TC0600010572.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171833.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171833.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171833.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171833.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171833.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171833.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171833.hg.1 TC0600010572.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600171836.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171836.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171836.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171836.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171836.hg.1 TC0600010572.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600171838.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171838.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171838.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171838.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171838.hg.1 TC0600010572.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600171834.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171834.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171834.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171834.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171834.hg.1 ENST00000473006.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171834.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171834.hg.1 TC0600010572.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600171837.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171837.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171837.hg.1 ENST00000259818.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600171837.hg.1 NM_178012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600171837.hg.1 TC0600010572.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600010567.hg.1 ENST00000404155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010567.hg.1 ENST00000404155.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600010567.hg.1 ENST00000404155.1 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 203030 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline