Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00608037_m1
          See other CDK6 GE Assays ›
          Gene Symbol:
          CDK6
          Gene Name:
          cyclin dependent kinase 6
          Gene Aliases:
          MCPH12, PLSTIRE
          Chromosome Location:
          Chr.7: 92604921 - 92836627 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00608037_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CDK6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145306.1 NP_001138778.1 4 - 5 1056 73
          NM_001259.6 NP_001250.1 4 - 5 951 73
          XM_006715835.2 XP_006715898.1 4 - 5 1176 73
          GenBank mRNA AK313491.1 - 4 - 5 994 73
          BC052264.1 - 4 - 5 932 6377987 73
          X66365.1 - 3 - 4 656 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700011785.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001603.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CDK6

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          positive regulation of cell-matrix adhesion
          type B pancreatic cell development
          protein phosphorylation
          cell cycle arrest
          Notch signaling pathway
          negative regulation of cell proliferation
          response to virus
          regulation of gene expression
          positive regulation of gene expression
          astrocyte development
          dentate gyrus development
          lateral ventricle development
          T cell differentiation in thymus
          gliogenesis
          cell dedifferentiation
          negative regulation of cell differentiation
          negative regulation of myeloid cell differentiation
          regulation of erythrocyte differentiation
          negative regulation of monocyte differentiation
          negative regulation of osteoblast differentiation
          negative regulation of cell cycle
          positive regulation of fibroblast proliferation
          generation of neurons
          negative regulation of epithelial cell proliferation
          cell division
          hematopoietic stem cell differentiation
          cellular response to phorbol 13-acetate 12-myristate
          regulation of cell motility
          negative regulation of cellular senescence
          cyclin-dependent protein serine/threonine kinase activity
          protein binding
          ATP binding
          cyclin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07024325.hg.1 TC07001603.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07024325.hg.1 ENST00000265734.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07024325.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07024325.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07024325.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07024326.hg.1 TC07001603.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07024326.hg.1 ENST00000265734.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07024326.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07024326.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07024326.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000491250.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 ENST00000491250.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay PSR0700184221.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184221.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184221.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184221.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184221.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay PSR0700184221.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184221.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184221.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184221.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184221.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay PSR0700184221.hg.1 TC0700011785.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700184222.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184222.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184222.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184222.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184222.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay PSR0700184222.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184222.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184222.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700184222.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700184222.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay PSR0700184222.hg.1 TC0700011785.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000265734.8 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000424848.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000467166.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000473078.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000491250.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 ENST00000491250.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001145306.1 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq
          Affymetrix Human Clariom D Assay TC0700011785.hg.1 NM_001259.7 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0