Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00610060_m1
          See other SFRP1 GE Assays ›
          Gene Symbol:
          SFRP1
          Gene Name:
          secreted frizzled related protein 1
          Gene Aliases:
          FRP, FRP-1, FRP1, FrzA, SARP2
          Chromosome Location:
          Chr.8: 41261957 - 41309471 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00610060_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SFRP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003012.4 NP_003003.3 2 - 3 934 130
          GenBank mRNA AF001900.1 - 2 - 3 921 130
          AF017987.1 - 2 - 3 924 130
          AF056087.1 - 2 - 3 921 130
          AK127331.1 - 2 - 3 347 130
          AK293090.1 - 3 - 4 747 130
          BC036503.1 - 2 - 3 941 5265277 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800010234.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08001166.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Human WNT Pathway 1700 Inventoried Human_Wnt Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SFRP1

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          ureteric bud development
          positive regulation of cell-matrix adhesion
          hematopoietic progenitor cell differentiation
          proteolysis
          G-protein coupled receptor signaling pathway
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          male gonad development
          female gonad development
          cellular response to starvation
          dorsal/ventral axis specification
          regulation of cell cycle process
          negative regulation of gene expression
          negative regulation of epithelial to mesenchymal transition
          neural crest cell fate commitment
          response to organic cyclic compound
          menstrual cycle phase
          positive regulation of Wnt signaling pathway
          negative regulation of Wnt signaling pathway
          negative regulation of ossification
          positive regulation of cell growth
          negative regulation of cell growth
          negative regulation of cell migration
          negative regulation of BMP signaling pathway
          negative regulation of osteoblast proliferation
          somatic stem cell population maintenance
          response to drug
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of JUN kinase activity
          positive regulation of GTPase activity
          cellular response to fibroblast growth factor stimulus
          stromal-epithelial cell signaling involved in prostate gland development
          negative regulation of B cell differentiation
          positive regulation of fat cell differentiation
          negative regulation of osteoblast differentiation
          negative regulation of osteoclast differentiation
          regulation of angiogenesis
          positive regulation of smoothened signaling pathway
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          negative regulation of insulin secretion
          negative regulation of bone remodeling
          negative regulation of fibroblast proliferation
          digestive tract morphogenesis
          positive regulation of epithelial cell proliferation
          negative regulation of epithelial cell proliferation
          negative regulation of peptidyl-tyrosine phosphorylation
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          canonical Wnt signaling pathway
          hematopoietic stem cell differentiation
          bone trabecula formation
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          regulation of branching involved in prostate gland morphogenesis
          negative regulation of androgen receptor signaling pathway
          cellular response to vitamin D
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to growth factor stimulus
          cellular response to prostaglandin E stimulus
          cellular response to estrogen stimulus
          cellular response to estradiol stimulus
          cellular response to hypoxia
          cellular response to X-ray
          cellular response to heparin
          dopaminergic neuron differentiation
          cellular response to transforming growth factor beta stimulus
          cellular response to BMP stimulus
          negative regulation of canonical Wnt signaling pathway
          planar cell polarity pathway involved in neural tube closure
          Wnt signaling pathway involved in somitogenesis
          convergent extension involved in somitogenesis
          positive regulation of canonical Wnt signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          regulation of dopaminergic neuron differentiation
          midbrain morphogenesis
          negative regulation of planar cell polarity pathway involved in axis elongation
          positive regulation of non-canonical Wnt signaling pathway
          negative regulation of Wnt signaling pathway involved in dorsal/ventral axis specification
          negative regulation of canonical Wnt signaling pathway involved in controlling type B pancreatic cell proliferation
          negative regulation of fibroblast apoptotic process
          positive regulation of fibroblast apoptotic process
          cysteine-type endopeptidase activity
          G-protein coupled receptor activity
          frizzled binding
          protein binding
          drug binding
          heparin binding
          Wnt-protein binding
          identical protein binding
          Wnt-activated receptor activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR08014981.hg.1 ENST00000220772.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014981.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014981.hg.1 ENST00000535263.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014981.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08014981.hg.1 TC08001166.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08014983.hg.1 ENST00000220772.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014983.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014983.hg.1 ENST00000535263.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014983.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08014983.hg.1 TC08001166.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08014984.hg.1 ENST00000220772.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014984.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014984.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08014984.hg.1 TC08001166.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08014988.hg.1 ENST00000220772.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014988.hg.1 ENST00000535263.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014988.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08014988.hg.1 TC08001166.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010234.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08014985.hg.1 ENST00000220772.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014985.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014985.hg.1 ENST00000535263.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014985.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08014985.hg.1 TC08001166.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08014989.hg.1 ENST00000220772.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014989.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08014989.hg.1 TC08001166.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08014990.hg.1 ENST00000220772.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014990.hg.1 ENST00000535263.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014990.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08014990.hg.1 TC08001166.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08014980.hg.1 ENST00000220772.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014980.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014980.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08014980.hg.1 TC08001166.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08014986.hg.1 ENST00000220772.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014986.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014986.hg.1 ENST00000535263.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014986.hg.1 NM_003012.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08014986.hg.1 TC08001166.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay TC0800010234.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800169644.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169644.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169644.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800169644.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169644.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169644.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800169644.hg.1 TC0800010234.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800169648.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169648.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800169648.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169648.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800169648.hg.1 TC0800010234.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800169643.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169643.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169643.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800169643.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169643.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169643.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800169643.hg.1 TC0800010234.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800169645.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169645.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169645.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800169645.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169645.hg.1 ENST00000379845.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169645.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800169645.hg.1 TC0800010234.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800169647.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169647.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800169647.hg.1 ENST00000220772.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0800169647.hg.1 NM_003012.4 Refseq
          Affymetrix Human Clariom D Assay PSR0800169647.hg.1 TC0800010234.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 143998 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline