Search
Search

C5AR1| Gene Symbol: C5AR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001736.3 | NP_001727.1 | 2 | 583 | 68 | |
| XM_005259190.4 | XP_005259247.1 | 2 | 556 | 68 | ||
| GenBank mRNA | BC008982.1 | - | 2 | 611 | 4177114 | 68 |
| BI916506.1 | - | 2 | 583 | 68 | ||
| BT007358.1 | - | 1 | 534 | 68 | ||
| M62505.1 | - | 2 | 558 | 68 | ||
| X57250.1 | - | 1 | 534 | 68 | ||
Set Membership: |
|