Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00708335_s1
          See other ERVV-1 GE Assays ›
          Gene Symbol:
          ERVV-1
          Gene Name:
          endogenous retrovirus group V member 1
          Gene Aliases:
          ENVV1, HERV-V1
          Chromosome Location:
          Chr.19: 53014091 - 53016580 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00708335_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ERVV-1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152473.2 NP_689686.2 1 1832 55
          GenBank mRNA AK056776.1 - 2 1557 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900008726.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000804.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ERVV-1

          Panther Classification:

          Molecular Function -

          viral or transposable element protein

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10996648 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10996647 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10996645 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10996644 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10996646 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10996643 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31637244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10996649 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P4466 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P293158 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 11171 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 29664 - -
          Agilent Human Genome CGH Microarray 44B A_14_P101354 - -
          Agilent Human Genome CGH Microarray 44A A_14_P101354 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P293158 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P4466 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P101354 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10996683 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10996684 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10996686 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10996688 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17157975 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10996685 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P10996687 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P101354 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P293158 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P4466 - -
          Agilent Human 1B Microarray G4111A A_32_P4466 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8180201 - -
          Affymetrix Human Gene 1.1 ST Array 8180201 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553255_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1900008726.hg.1 ENST00000602168.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008726.hg.1 ENST00000602168.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008726.hg.1 ENST00000602168.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008726.hg.1 NM_152473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008726.hg.1 NM_152473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008726.hg.1 NM_152473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19011311.hg.1 NM_152473.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19011311.hg.1 TC19000804.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1900168209.hg.1 ENST00000602168.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900168209.hg.1 NM_152473.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900168209.hg.1 TC1900008726.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900168209.hg.1 ENST00000602168.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900168209.hg.1 NM_152473.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900168208.hg.1 ENST00000602168.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900168208.hg.1 NM_152473.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900168208.hg.1 TC1900008726.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900168208.hg.1 ENST00000602168.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900168208.hg.1 NM_152473.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900168210.hg.1 NM_152473.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900168210.hg.1 TC1900008726.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900168210.hg.1 NM_152473.2 Refseq
          Affymetrix Human Clariom D Assay TC1900008726.hg.1 ENST00000602168.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008726.hg.1 ENST00000602168.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008726.hg.1 ENST00000602168.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008726.hg.1 NM_152473.2 Refseq
          Affymetrix Human Clariom D Assay TC1900008726.hg.1 NM_152473.2 Refseq
          Affymetrix Human Clariom D Assay TC1900008726.hg.1 NM_152473.2 Refseq
          Human Genome U133 Plus 2.0 Array 1553255_at - -
          Human Genome U133 Plus 2.0 Array 1553255_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_152473_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553255_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553255_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553255_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553255_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553255_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553255_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553255_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553255_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553255_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553255_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553255_at11 - -
          Human Genome U95 Set 44994_PM_at - -
          Human Genome U95 Set 44994_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 220282 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1780637 NM_152473.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1780637 AK056776.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1780637 NM_152473.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1780637 AK056776.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1780637 NM_152473.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1780637 AK056776.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_22748988-S NM_152473.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_22748988-S AK056776.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_4844-S - -
          Illumina human-6 v1.0 expression beadchip 0005910706 - -
          Illumina humanRef-8 v1.0 expression beadchip 0005910706 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1780637 NM_152473.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1780637 AK056776.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1780637 NM_152473.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1780637 AK056776.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 2350246 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_4844 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1780637 NM_152473.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1780637 AK056776.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1780637 NM_152473.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1780637 AK056776.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1780637 NM_152473.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1780637 AK056776.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline