Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00747223_g1
          See other HBB GE Assays ›
          Gene Symbol:
          HBB
          Gene Name:
          hemoglobin subunit beta
          Gene Aliases:
          CD113t-C, beta-globin
          Chromosome Location:
          Chr.11: 5225466 - 5227071 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00747223_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HBB
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000518.4 NP_000509.1 1 - 2 145 106
          GenBank mRNA AF117710.1 - 1 - 2 125 106
          AF181989.1 - 1 - 2 128 106
          AF349114.1 - 1 - 2 130 106
          AK311825.1 - 1 - 2 145 106
          AY136510.1 - 1 - 2 95 106
          CR536530.1 - 1 - 2 95 106
          CR541913.1 - 1 - 2 95 106
          EU694432.1 - 1 - 2 142 103
          M25113.1 - 1 - 2 95 106
          V00497.1 - 1 - 2 145 106
          V00500.1 - 1 - 2 95 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100009917.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001330.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Human Stem Cell Pluripotency Inventoried Human_Stem_Cell Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HBB

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor-mediated endocytosis
          blood coagulation
          regulation of blood pressure
          positive regulation of cell death
          oxygen transport
          bicarbonate transport
          nitric oxide transport
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          positive regulation of nitric oxide biosynthetic process
          regulation of blood vessel size
          protein heterooligomerization
          renal absorption
          platelet aggregation
          cellular oxidant detoxification
          peroxidase activity
          oxygen transporter activity
          iron ion binding
          protein binding
          oxygen binding
          heme binding
          hemoglobin binding
          haptoglobin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016741.hg.1 ENST00000335295.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016741.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016741.hg.1 TC11001330.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016738.hg.1 ENST00000335295.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016738.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016738.hg.1 TC11001330.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11016737.hg.1 ENST00000335295.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016737.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016737.hg.1 TC11001330.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11016740.hg.1 ENST00000335295.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016740.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016740.hg.1 TC11001330.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100174842.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174842.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174842.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174842.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174842.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100174842.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174842.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174842.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174842.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174842.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100174842.hg.1 TC1100009917.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100174843.hg.1 TC1100009917.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174846.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174846.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174846.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174846.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174846.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100174846.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174846.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174846.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174846.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174846.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100174846.hg.1 TC1100009917.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100174848.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174848.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174848.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174848.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100174848.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174848.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174848.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174848.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100174848.hg.1 TC1100009917.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline