Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00747483_s1
          See other H3C3 GE Assays ›
          Gene Symbol:
          H3C3
          Gene Name:
          H3 clustered histone 3
          Gene Aliases:
          H3.1, H3/c, H3C1, H3C10, H3C11, H3C12, H3C2, H3C4, H3C6, H3C7, H3C8, H3FC, HIST1H3C
          Chromosome Location:
          Chr.6: 26045384 - 26045869 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00747483_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H3C3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003531.3 NP_003522.1 1 448 104
          GenBank mRNA BC127610.1 - 1 421 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600007265.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000164.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Human Interferon Pathway 1700 Inventoried Human_Interferon_Pathway Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H3C3

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          nucleosome assembly
          gene expression
          telomere organization
          regulation of gene expression, epigenetic
          DNA binding
          protein binding
          structural constituent of chromatin
          cadherin binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624487 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24812722 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624482 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624491 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24812717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24812708 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624478 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624480 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624494 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24812725 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24812718 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624488 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P133814 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P133814 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 34922 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 49649 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P31700989 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P133814 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P133814 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624482 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624495 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624497 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624480 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812708 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624494 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624478 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624488 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624475 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624487 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812722 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812717 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624476 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624480 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812731 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624497 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624475 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624495 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812708 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812718 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624487 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624488 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624482 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624478 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624494 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812722 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P133814 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624482 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624494 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624497 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624490 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624485 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624495 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624492 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624487 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04624481 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04624479 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04624482 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P133814 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 2845 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624453 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624472 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624485 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624476 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624497 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624450 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624510 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624464 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624505 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624516 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624494 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624444 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624462 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624467 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624466 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624514 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624492 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624478 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624487 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624517 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624512 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624482 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624502 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624475 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624521 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624519 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15793600 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624470 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624490 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624508 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_003531_at - -
          Affymetrix Human Gene 1.0 ST Array 8117340 - -
          Affymetrix Human Gene 1.0 ST Array 8117341 - -
          Affymetrix Human Gene 1.1 ST Array 8117339 - -
          Affymetrix HT HG-U133+ PM Array Plate 208577_PM_at - -
          Affymetrix Human Genome U219 Array 11738783_at - -
          Affymetrix Human Genome U219 Array 11738783_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06001667.hg.1 ENST00000540144.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06001667.hg.1 NM_003531.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06001667.hg.1 TC06000164.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600149264.hg.1 NM_003531.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149264.hg.1 NM_003531.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149264.hg.1 TC0600007265.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 208496_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 214472_PM_at - -
          Human Genome U133 Plus 2.0 Array 208506_at - -
          Human Genome U133 Plus 2.0 Array 208496_x_at - -
          Human Genome U133 Plus 2.0 Array 208576_s_at - -
          Human Genome U133 Plus 2.0 Array 208575_at - -
          Human Genome U133 Plus 2.0 Array 208577_at - -
          Human Genome U133 Plus 2.0 Array 208576_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 208575_PM_at - -
          Human Genome U133 Plus 2.0 Array 214616_PM_at - -
          Human Genome U133 Plus 2.0 Array 214616_at - -
          Human Genome U133 Plus 2.0 Array 208506_PM_at - -
          Human Genome U133 Plus 2.0 Array 214472_at - -
          Human Genome U133 Plus 2.0 Array 208577_PM_at - -
          Human Genome U133A 2.0 Array 208575_PM_at - -
          Human Genome U133A 2.0 Array 208496_PM_x_at - -
          Human Genome U133A 2.0 Array 208576_PM_s_at - -
          Human Genome U133A 2.0 Array 208577_at - -
          Human Genome U133A 2.0 Array 214616_PM_at - -
          Human Genome U133A 2.0 Array 208506_PM_at - -
          Human Genome U133A 2.0 Array 208576_s_at - -
          Human Genome U133A 2.0 Array 208496_x_at - -
          Human Genome U133A 2.0 Array 208506_at - -
          Human Genome U133A 2.0 Array 208575_at - -
          Human Genome U133A 2.0 Array 214472_at - -
          Human Genome U133A 2.0 Array 214616_at - -
          Human Genome U133A 2.0 Array 208577_PM_at - -
          Human Genome U133A 2.0 Array 214472_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_003531_at - -
          AffymetrixPathwork Diagnostics Pathchip pw536976 - -
          Human Genome U95 Set 35598_at - -
          Human Genome U95 Set 35598_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208577_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208577_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208577_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208577_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208577_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208577_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208577_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208577_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208577_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208577_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208577_at8 - -
          Human Genome U133 Set 208506_PM_at - -
          Human Genome U133 Set 208496_PM_x_at - -
          Human Genome U133 Set 208496_x_at - -
          Human Genome U133 Set 208506_at - -
          Human Genome U133 Set 214472_at - -
          Human Genome U133 Set 214616_PM_at - -
          Human Genome U133 Set 214472_PM_at - -
          Human Genome U133 Set 214616_at - -
          Human Genome U133 Set 208576_s_at - -
          Human Genome U133 Set 208575_PM_at - -
          Human Genome U133 Set 208576_PM_s_at - -
          Human Genome U133 Set 208575_at - -
          Human Genome U133 Set 208577_at - -
          Human Genome U133 Set 208577_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031094 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031094 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 163028 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1712184 NM_003531.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1712184 BC058834.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1712184 BC127610.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1712184 NM_003531.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1712184 BC058834.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1712184 BC127610.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1712184 NM_003531.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1712184 BC058834.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1712184 BC127610.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1712184 NM_003531.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1712184 BC058834.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1712184 BC127610.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1712184 NM_003531.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1712184 BC058834.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1712184 BC127610.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21071022-S NM_003531.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21071022-S BC058834.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21071022-S BC127610.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_23964-S - -
          Illumina human-6 v1.0 expression beadchip 0001740113 - -
          Illumina humanRef-8 v1.0 expression beadchip 0001740113 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1712184 NM_003531.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1712184 BC058834.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1712184 BC127610.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1712184 NM_003531.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1712184 BC058834.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1712184 BC127610.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 4120671 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_23964 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1712184 NM_003531.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1712184 BC058834.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1712184 BC127610.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1712184 NM_003531.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1712184 BC058834.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1712184 BC127610.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1712184 NM_003531.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1712184 BC058834.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1712184 BC127610.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1712184 NM_003531.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1712184 BC058834.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1712184 BC127610.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1712184 NM_003531.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1712184 BC058834.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1712184 BC127610.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline