Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00845911_m1
          See other LOC339059 GE Assays ›
          Gene Symbol:
          LOC339059
          Gene Name:
          uncharacterized LOC339059
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 88742767 - 88745748 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00845911_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC339059
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_120387.1 2 - 3 1474 75
          GenBank mRNA AK097767.1 - 2 - 3 1474 75
          BC111769.1 - 1 - 2 1521 40025355 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600008840.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724951 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179526 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179553 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179540 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724897 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179544 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179547 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179510 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179513 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724940 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179573 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179531 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179542 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179537 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724920 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724905 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16928209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179568 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16928190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179528 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724911 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179518 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179562 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179509 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179549 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724950 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179539 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724976 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30724900 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179564 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179541 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10179529 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 39500 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 45403 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3424848 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P898690 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 122940 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P122927 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7997833 - -
          Affymetrix Human Gene 1.0 ST Array 7997834 - -
          Affymetrix Human Gene 1.0 ST Array 7997835 - -
          Affymetrix Human Gene 1.0 ST Array 7997836 - -
          Affymetrix Human Gene 1.0 ST Array 7997837 - -
          Affymetrix Human Gene 1.0 ST Array 7997838 - -
          Affymetrix Human Gene 1.1 ST Array 7997832 - -
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165880.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165880.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165880.hg.1 NR_120387.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165880.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165880.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165880.hg.1 NR_120387.1 Refseq
          Affymetrix Human Clariom D Assay PSR1600165880.hg.1 TC1600008840.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600165877.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165877.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165877.hg.1 TC1600008840.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000333666.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 ENST00000567588.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 NR_120387.1 Refseq
          Affymetrix Human Clariom D Assay TC1600008840.hg.1 NR_120387.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 384551 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1764029 BC111769.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1764029 AK097767.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3307134 BC111769.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3307134 AK097767.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1764029 BC111769.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1764029 AK097767.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1764029 BC111769.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1764029 AK097767.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1764029 BC111769.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1764029 AK097767.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1764029 BC111769.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1764029 AK097767.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1764029 BC111769.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1764029 AK097767.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1764029 BC111769.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1764029 AK097767.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1764029 BC111769.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1764029 AK097767.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1764029 BC111769.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1764029 AK097767.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline