Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00870554_g1
          See other IGLJ3 GE Assays ›
          Gene Symbol:
          IGLJ3 CYAT1
          Gene Name:
          immunoglobulin lambda joining 3
          immunoglobulin lambda light chain-like
          Gene Aliases:
          J3
          -
          Chromosome Location:
          Chr.22: 22904988 - 22905025 on Build GRCh38
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs00870554_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IGLJ3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA X57817.1 - 2 237 136
          Gene Symbol: CYAT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC093098.1 - 2 291 30325463 136

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          immunoglobulin

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P605563 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3395675 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3336822 - -
          Agilent Human 2 cDNA Microarray (G4101A) 3054669 - -
          Agilent Human 2 cDNA Microarray (G4101A) 3054669 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7981731 - -
          Affymetrix Human Gene 1.1 ST Array 7981730 - -
          Affymetrix HT HG-U133+ PM Array Plate 211881_PM_x_at - -
          Affymetrix HT HG-U133+ PM Array Plate 214677_PM_x_at - -
          Affymetrix HT HG-U133+ PM Array Plate 234884_PM_x_at - -
          Affymetrix HT HG-U133+ PM Array Plate 211798_PM_x_at - -
          Affymetrix HT HG-U133+ PM Array Plate 234366_PM_x_at - -
          Affymetrix Human Transcriptome Array 2.0 TC2200009210.hg.1 ENST00000390301.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009210.hg.1 ENST00000390301.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009209.hg.1 ENST00000390299.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009209.hg.1 ENST00000390299.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006817.hg.1 ENST00000517690.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009208.hg.1 ENST00000390295.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009208.hg.1 ENST00000390295.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009208.hg.1 ENST00000390296.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009208.hg.1 ENST00000390296.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009208.hg.1 ENST00000390297.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009208.hg.1 ENST00000390297.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009208.hg.1 ENST00000390297.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009208.hg.1 ENST00000390298.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009208.hg.1 ENST00000390298.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009208.hg.1 ENST00000443400.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006771.hg.1 ENST00000390291.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200006771.hg.1 ENST00000390291.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009222.hg.1 ENST00000390325.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009207.hg.1 ENST00000390294.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200009207.hg.1 ENST00000390294.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200009222.hg.1 ENST00000390325.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200009209.hg.1 ENST00000390299.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200009209.hg.1 ENST00000390299.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200006817.hg.1 ENST00000517690.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200009207.hg.1 ENST00000390294.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200009207.hg.1 ENST00000390294.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200009210.hg.1 ENST00000390301.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009210.hg.1 ENST00000390301.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009208.hg.1 ENST00000390295.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009208.hg.1 ENST00000390295.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009208.hg.1 ENST00000390296.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200009208.hg.1 ENST00000390296.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200009208.hg.1 ENST00000390297.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009208.hg.1 ENST00000390297.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009208.hg.1 ENST00000390297.3 Ensembl
          Affymetrix Human Clariom D Assay TC2200009208.hg.1 ENST00000390298.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200009208.hg.1 ENST00000390298.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200009208.hg.1 ENST00000443400.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200006771.hg.1 ENST00000390291.2 Ensembl
          Affymetrix Human Clariom D Assay TC2200006771.hg.1 ENST00000390291.2 Ensembl
          Human Genome U133 Plus 2.0 Array 216852_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 216853_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 216851_at - -
          Human Genome U133 Plus 2.0 Array 211798_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 211881_x_at - -
          Human Genome U133 Plus 2.0 Array 211881_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 203174_s_at - -
          Human Genome U133 Plus 2.0 Array 211798_x_at - -
          Human Genome U133 Plus 2.0 Array 215984_s_at - -
          Human Genome U133 Plus 2.0 Array 214677_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 216851_PM_at - -
          Human Genome U133 Plus 2.0 Array 216853_x_at - -
          Human Genome U133 Plus 2.0 Array 215984_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 216852_x_at - -
          Human Genome U133 Plus 2.0 Array 214677_x_at - -
          Human Genome U133 Plus 2.0 Array 203174_PM_s_at - -
          Human Genome U133A 2.0 Array 203174_PM_s_at - -
          Human Genome U133A 2.0 Array 215984_s_at - -
          Human Genome U133A 2.0 Array 214677_PM_x_at - -
          Human Genome U133A 2.0 Array 211881_PM_x_at - -
          Human Genome U133A 2.0 Array 211881_x_at - -
          Human Genome U133A 2.0 Array 215984_PM_s_at - -
          Human Genome U133A 2.0 Array 203174_s_at - -
          Human Genome U133A 2.0 Array 214677_x_at - -
          Human Genome U133A 2.0 Array 211798_x_at - -
          Human Genome U133A 2.0 Array 211798_PM_x_at - -
          AffymetrixPathwork Diagnostics Pathchip pw585174 - -
          AffymetrixPathwork Diagnostics Pathchip pw502325 - -
          AffymetrixPathwork Diagnostics Pathchip pw567208 - -
          Human Genome U95 Set 35765_at - -
          Human Genome U95 Set 35765_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211881_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211798_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211798_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211881_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211881_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214677_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214677_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211798_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214677_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214677_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214677_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211798_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211881_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211798_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211798_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211798_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211881_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211798_x_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214677_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214677_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211881_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214677_x_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211798_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211881_x_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214677_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211881_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214677_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 214677_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211881_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211881_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211798_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211881_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 211798_x_at6 - -
          Human Genome U95 Set 73327_r_at - -
          Human Genome U95 Set 73327_PM_r_at - -
          Human Genome U133 Set 203174_s_at - -
          Human Genome U133 Set 215984_PM_s_at - -
          Human Genome U133 Set 214677_PM_x_at - -
          Human Genome U133 Set 211798_PM_x_at - -
          Human Genome U133 Set 211881_x_at - -
          Human Genome U133 Set 211798_x_at - -
          Human Genome U133 Set 203174_PM_s_at - -
          Human Genome U133 Set 211881_PM_x_at - -
          Human Genome U133 Set 214677_x_at - -
          Human Genome U133 Set 215984_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 172692 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-v472f:80/100.66.78.195:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline