Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00885169_s1
          See other TCEAL6 GE Assays ›
          Gene Symbol:
          TCEAL6
          Gene Name:
          transcription elongation factor A like 6
          Gene Aliases:
          Tceal3, WEX2
          Chromosome Location:
          Chr.X: 102139961 - 102142416 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00885169_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TCEAL6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001006938.2 NP_001006939.2 3 1412 144

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TCEAL6

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          WW domain binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582688 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582662 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17380848 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11854372 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582649 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11854371 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582689 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17380835 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11854376 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582651 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11854358 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11854354 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11854365 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582694 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11854360 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11854362 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11854384 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11854361 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11854370 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582648 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582685 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582655 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582653 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32582652 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11854374 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 153456 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 116407 AB209562.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 116407 AK096385.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 116407 BC013220.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 116407 NM_002140.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 116407 NM_031262.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 116407 NM_031263.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 116407 X72727.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 284357 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P192545 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3356910 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P192545 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 5071 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 23747 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 54252 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P41744401 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P192545 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3356910 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P192545 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3356910 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854361 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582671 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17380848 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582663 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854372 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582685 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582660 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854382 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17380835 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854374 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582689 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854362 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582687 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854370 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854383 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582688 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582694 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854376 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854371 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582655 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582662 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582671 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854372 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582688 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582655 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854376 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582687 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17380848 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582669 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854381 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582662 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582685 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582689 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854361 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854371 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582660 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854370 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854382 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17380835 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854383 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854374 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582694 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11854362 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P32582663 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P192545 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11854367 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11854372 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11854376 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11854369 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11854374 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11854371 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P11854370 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P17380852 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P192545 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 122334 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 4082 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41744401 - -
          Agilent Human 1B Microarray G4111A A_32_P192547 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854371 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854374 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854364 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854341 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854354 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854378 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854379 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854350 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854343 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854384 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854362 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854346 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854369 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854382 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854370 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854338 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854376 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854349 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854356 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854381 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854340 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854380 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854337 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854359 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854367 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854366 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11854352 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P41744401 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001006938_at - -
          Affymetrix Human Gene 1.0 ST Array 8174138 - -
          Affymetrix Human Gene 1.0 ST Array 8174137 - -
          Affymetrix Human Gene 1.0 ST Array 8174139 - -
          Affymetrix Human Gene 1.0 ST Array 8174136 - -
          Affymetrix Human Gene 1.0 ST Array 8174135 - -
          Affymetrix Human Gene 1.1 ST Array 8174134 - -
          Affymetrix Human Genome U219 Array 11737036_at - -
          Affymetrix Human Genome U219 Array 11737036_PM_at - -
          Affymetrix Human Genome U219 Array 11737037_x_at - -
          Affymetrix Human Genome U219 Array 11737037_PM_x_at - -
          Human Genome U95 Set 71101_PM_at - -
          Human Genome U95 Set 71101_at - -
          Human Genome U95 Set 87383_at - -
          Human Genome U95 Set 87383_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040204 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A040204 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 152643 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3251393 NM_001006938.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1729165 NM_001006938.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1729165 BC071675.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1697473 - -
          HumanHT-12 V4.0 expression beadchip ILMN_1729165 NM_001006938.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1729165 BC071675.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3251393 NM_001006938.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1729165 NM_001006938.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1729165 BC071675.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3251393 NM_001006938.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1729165 NM_001006938.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1729165 BC071675.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3251393 NM_001006938.2 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_5218-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1697473 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1697473 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 5910673 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_5218 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1729165 NM_001006938.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1729165 BC071675.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1729165 NM_001006938.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1729165 BC071675.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1729165 NM_001006938.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1729165 BC071675.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1697473 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1729165 NM_001006938.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1729165 BC071675.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline