Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00898625_g1
          See other YBX1 GE Assays ›
          Gene Symbol:
          YBX1
          Gene Name:
          Y-box binding protein 1
          Gene Aliases:
          BP-8, CBF-A, CSDA2, CSDB, DBPB, EFI-A, MDR-NF1, NSEP-1, NSEP1, YB-1, YB1
          Chromosome Location:
          Chr.1: 42682235 - 42703803 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00898625_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: YBX1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004559.4 NP_004550.2 5 - 6 990 74
          NR_132737.1 4 - 5 808 74
          GenBank mRNA AK309996.1 - 5 - 6 627 74
          BC000064.2 - 5 - 6 777 74
          BC002411.2 - 5 - 6 773 2961659 74
          BC010430.1 - 5 - 6 772 3506647 74
          BC013838.1 - 4 - 5 808 74
          BC015208.1 - 5 - 6 722 3914485 74
          BC018393.1 - 5 - 6 757 74
          BC020164.1 - 1 231 74
          BC038384.1 - 5 - 6 779 3449398 74
          BC065571.1 - 5 - 6 797 6171221 74
          BC070084.1 - 5 - 6 797 5272578 74
          BC071708.1 - 5 - 6 782 4361396 74
          BC072014.1 - 5 - 6 782 74
          BC090038.1 - 5 - 6 782 6526492 74
          BC098435.1 - 5 - 6 788 4859074 74
          BC106045.1 - 5 - 6 784 6062615 74
          J03827.1 - 5 - 6 785 74
          L28809.1 - 5 - 6 740 74
          M24070.1 - 5 - 6 779 74
          M83234.1 - 1 881 74
          M85234.1 - 1 887 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100008025.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000531.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene YBX1

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of cell division
          CRD-mediated mRNA stabilization
          protein localization to cytoplasmic stress granule
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          RNA binding
          protein binding
          poly(A) RNA binding
          GTPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01008386.hg.1 TC01000531.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01008386.hg.1 ENST00000318612.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008386.hg.1 ENST00000321358.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008386.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01008383.hg.1 TC01000531.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01008383.hg.1 ENST00000318612.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008383.hg.1 ENST00000321358.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01008383.hg.1 NM_004559.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700012164.hg.1 ENST00000489612.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012164.hg.1 ENST00000489612.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700012164.hg.1 ENST00000489612.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0100008025.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay TC0700012164.hg.1 ENST00000489612.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012164.hg.1 ENST00000489612.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700012164.hg.1 ENST00000489612.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 ENST00000467957.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100156583.hg.1 TC0100008025.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100156584.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156584.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156584.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156584.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156584.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100156584.hg.1 ENST00000321358.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156584.hg.1 ENST00000332220.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156584.hg.1 ENST00000436427.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156584.hg.1 ENST00000616651.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100156584.hg.1 NM_004559.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100156584.hg.1 TC0100008025.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0