Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00900828_g1
          See other IL23A GE Assays ›
          Gene Symbol:
          IL23A
          Gene Name:
          interleukin 23 subunit alpha
          Gene Aliases:
          IL-23, IL-23A, IL23P19, P19, SGRF
          Chromosome Location:
          Chr.12: 56334159 - 56340410 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00900828_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IL23A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016584.2 NP_057668.1 2 - 3 429 81
          XM_011538477.2 XP_011536779.1 4 - 5 616 81
          GenBank mRNA AB030000.1 - 2 - 3 406 81
          AF301620.1 - 2 - 3 413 81
          AY359083.1 - 2 - 3 433 81
          BC066267.1 - 2 - 3 296 6971848 81
          BC066268.1 - 2 - 3 296 6971849 81
          BC066269.1 - 2 - 3 296 6971851 81
          BC067511.1 - 2 - 3 296 6971850 81
          BC067512.1 - 2 - 3 296 6971852 81
          BC067513.1 - 2 - 3 296 6971853 81
          BM842783.1 - 2 - 3 429 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200007838.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000512.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IL23A

          Panther Classification:

          Molecular Function -

          cytokine interleukin superfamily intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          positive regulation of defense response to virus by host
          positive regulation of T-helper 1 type immune response
          inflammatory response
          positive regulation of activation of JAK2 kinase activity
          negative regulation of interleukin-10 production
          positive regulation of granulocyte macrophage colony-stimulating factor production
          positive regulation of interferon-gamma production
          positive regulation of interleukin-10 production
          positive regulation of interleukin-12 production
          positive regulation of interleukin-17 production
          positive regulation of tumor necrosis factor production
          positive regulation of natural killer cell activation
          positive regulation of natural killer cell proliferation
          positive regulation of tissue remodeling
          T cell proliferation
          positive regulation of T cell proliferation
          positive regulation of activated T cell proliferation
          positive regulation of NF-kappaB import into nucleus
          regulation of tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of tyrosine phosphorylation of Stat4 protein
          positive regulation of tyrosine phosphorylation of Stat5 protein
          positive regulation of memory T cell differentiation
          innate immune response
          positive regulation of osteoclast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          tissue remodeling
          positive regulation of inflammatory response
          defense response to Gram-negative bacterium
          positive regulation of NK T cell activation
          positive regulation of NK T cell proliferation
          defense response to virus
          positive regulation of neutrophil chemotaxis
          positive regulation of T-helper 17 type immune response
          positive regulation of T-helper 17 cell lineage commitment
          cytokine activity
          protein binding
          interleukin-23 receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12006715.hg.1 ENST00000228534.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12006715.hg.1 NM_016584.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12006715.hg.1 TC12000512.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Clariom D Assay TC1200007838.hg.1 NM_016584.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200156959.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200156959.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200156959.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200156959.hg.1 NM_016584.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200156959.hg.1 ENST00000228534.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200156959.hg.1 ENST00000619177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200156959.hg.1 ENST00000622119.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200156959.hg.1 NM_016584.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200156959.hg.1 TC1200007838.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0