Search
Search

FUBP1| Gene Symbol: FUBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130152.1 | 5 - 6 | 541 | 105 | ||
| XM_011542392.2 | XP_011540694.1 | 6 - 7 | 509 | 105 | ||
| XM_017002741.1 | XP_016858230.1 | 6 - 7 | 493 | 105 | ||
| XM_017002742.1 | XP_016858231.1 | 6 - 7 | 509 | 105 | ||
| XM_017002743.1 | XP_016858232.1 | 5 - 6 | 450 | 105 | ||
| XM_017002744.1 | XP_016858233.1 | 5 - 6 | 450 | 105 | ||
| GenBank mRNA | AK301581.1 | - | 5 - 6 | 409 | 105 | |
| AK303568.1 | - | 5 - 6 | 408 | 105 | ||
| BC017247.2 | - | 5 - 6 | 405 | 4891583 | 105 | |
Set Membership: |
|