Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00962991_m1
          See other SHISA8 GE Assays ›
          Gene Symbol:
          SHISA8
          Gene Name:
          shisa family member 8
          Gene Aliases:
          C22orf17, Orf26
          Chromosome Location:
          Chr.22: 41907180 - 41915093 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00962991_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SHISA8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001207020.1 NP_001193949.1 1 - 2 528 142
          XM_006724254.3 XP_006724317.1 1 - 2 954 142
          XM_006724255.2 XP_006724318.1 1 - 2 954 142
          XM_006724256.3 XP_006724319.1 1 - 2 611 142
          GenBank mRNA AI763350.1 - 1 - 2 427 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200008845.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000834.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Homo sapiens 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          regulation of short-term neuronal synaptic plasticity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3222932 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 244467_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR22014092.hg.1 ENST00000457093.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22014092.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22014092.hg.1 TC22000834.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22014091.hg.1 ENST00000457093.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22014091.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22014091.hg.1 TC22000834.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22014090.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22014090.hg.1 TC22000834.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22014089.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22014089.hg.1 TC22000834.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 ENST00000457093.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 ENST00000457093.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22014093.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22014093.hg.1 TC22000834.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22014088.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22014088.hg.1 TC22000834.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 ENST00000457093.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 ENST00000457093.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay TC2200008845.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164080.hg.1 TC2200008845.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200164080.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164080.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164080.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164080.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164079.hg.1 TC2200008845.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200164079.hg.1 ENST00000457093.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164079.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164079.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164079.hg.1 ENST00000457093.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164079.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164079.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164077.hg.1 TC2200008845.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200164077.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164077.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164077.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164077.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164078.hg.1 TC2200008845.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200164078.hg.1 ENST00000457093.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164078.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164078.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164078.hg.1 ENST00000457093.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164078.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164078.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164075.hg.1 TC2200008845.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200164075.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164075.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164075.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164075.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164074.hg.1 TC2200008845.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200164074.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164074.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164074.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164074.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164076.hg.1 TC2200008845.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200164076.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164076.hg.1 NM_001207020.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200164076.hg.1 ENST00000621082.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200164076.hg.1 NM_001207020.2 Refseq
          Human Genome U133 Plus 2.0 Array 244467_at - -
          Human Genome U133 Plus 2.0 Array 244467_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244467_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244467_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244467_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244467_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244467_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244467_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244467_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244467_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244467_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244467_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 244467_at4 - -
          Human Genome U133 Set 244467_PM_at - -
          Human Genome U133 Set 244467_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3L0003232 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3L0003232 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 413070 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1672074 NM_001207020.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1672074 CR748983.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1672074 NM_001207020.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1672074 CR748983.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1672074 NM_001207020.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1672074 CR748983.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1672074 NM_001207020.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1672074 CR748983.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline