Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00966296_m1
          See other NF2 GE Assays ›
          Gene Symbol:
          NF2
          Gene Name:
          neurofibromin 2
          Gene Aliases:
          ACN, BANF, SCH
          Chromosome Location:
          Chr.22: 29603556 - 29698600 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00966296_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NF2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181832.2 NP_861970.1 16 - 17 2240 89
          GenBank mRNA AF369668.1 - 16 - 17 1797 89
          L28839.1 - 1 - 2 66 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200007027.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22000199.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NF2

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mesoderm formation
          negative regulation of cell-matrix adhesion
          negative regulation of protein kinase activity
          ectoderm development
          negative regulation of DNA replication
          negative regulation of cell proliferation
          Schwann cell proliferation
          regulation of gliogenesis
          hippocampus development
          negative regulation of cell-cell adhesion
          actin cytoskeleton organization
          negative regulation of cell growth
          negative regulation of cell migration
          regulation of protein stability
          regulation of hippo signaling
          odontogenesis of dentin-containing tooth
          negative regulation of tyrosine phosphorylation of Stat3 protein
          negative regulation of tyrosine phosphorylation of Stat5 protein
          negative regulation of MAPK cascade
          cell-cell junction organization
          positive regulation of cell differentiation
          negative regulation of JAK-STAT cascade
          positive regulation of stress fiber assembly
          lens fiber cell differentiation
          regulation of stem cell proliferation
          regulation of protein localization to nucleus
          regulation of neural precursor cell proliferation
          actin binding
          integrin binding
          protein binding
          beta-catenin binding
          protein domain specific binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 ENST00000338641.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 ENST00000347330.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 ENST00000353887.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 ENST00000361166.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 ENST00000361452.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 ENST00000361676.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 ENST00000397789.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 ENST00000397822.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 ENST00000413209.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 NM_000268.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 NM_016418.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 NM_181828.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 NM_181829.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 NM_181830.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 NM_181832.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 NM_181833.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22003724.hg.1 TC22000199.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 TC2200007027.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000338641.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000353887.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000361166.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000361452.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000361676.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000397789.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000413209.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000432151.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_000268.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_016418.5 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_181828.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_181829.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_181830.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_181832.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_181833.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000338641.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000353887.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000361166.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000361452.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000361676.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000397789.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000413209.6 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 ENST00000432151.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_000268.3 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_016418.5 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_181828.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_181829.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_181830.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_181832.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200149136.hg.1 NM_181833.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0