Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00967014_m1
          See other SNAPC4 GE Assays ›
          Gene Symbol:
          SNAPC4
          Gene Name:
          small nuclear RNA activating complex polypeptide 4
          Gene Aliases:
          NEDRSO, PTFalpha, SNAP190
          Chromosome Location:
          Chr.9: 136375571 - 136400170 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00967014_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNAPC4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001394201.1 NP_001381130.1 22 - 23 4333 60
          NM_001394202.1 NP_001381131.1 22 - 23 4249 60
          NM_001394203.1 NP_001381132.1 22 - 23 4245 60
          NM_003086.4 NP_003077.2 22 - 23 4329 60
          XM_006717242.5 XP_006717305.1 22 - 23 5510 60
          XM_006717244.3 XP_006717307.1 7 - 8 2587 60
          XM_047423780.1 XP_047279736.1 22 - 23 5426 60
          GenBank mRNA AF032387.1 - 21 - 22 4658 60
          BF339801.1 - 1 - 2 153 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900011976.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09001729.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SNAPC4

          Gene Ontology Categories:

          Function(s) Process(es)

          snRNA transcription from RNA polymerase II promoter
          snRNA transcription from RNA polymerase III promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcription factor activity, core RNA polymerase III binding
          RNA polymerase III type 3 promoter sequence-specific DNA binding
          DNA binding
          protein binding
          obsolete general RNA polymerase II transcription factor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09022113.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09022113.hg.1 NM_003086.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09022113.hg.1 TC09001729.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0900181996.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900181996.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900181996.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0900181996.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900181996.hg.1 TC0900011976.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 ENST00000298532.2 Ensembl
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq
          Affymetrix Human Clariom D Assay TC0900011976.hg.1 NM_003086.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0