Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00982312_m1
          See other MYLIP GE Assays ›
          Gene Symbol:
          MYLIP
          Gene Name:
          myosin regulatory light chain interacting protein
          Gene Aliases:
          IDOL, MIR
          Chromosome Location:
          Chr.6: 16129086 - 16163887 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00982312_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MYLIP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001436627.1 3 - 4 696 72
          NM_013262.4 NP_037394.2 3 - 4 696 72
          XM_005249032.2 XP_005249089.1 3 - 4 696 72
          XM_005249033.4 XP_005249090.1 2 - 3 505 72
          XM_017010789.2 XP_016866278.1 3 - 4 696 72
          XM_047418683.1 XP_047274639.1 3 - 4 696 72
          GenBank mRNA AF006003.1 - 3 - 4 609 72
          AF187016.2 - 3 - 4 633 72
          AF212221.1 - 2 - 3 420 72
          AF258586.1 - 3 - 4 673 72
          AK074391.1 - 3 - 4 696 72
          AK314648.1 - 3 - 4 696 72
          BC002860.2 - 3 - 4 677 72
          BT007055.1 - 3 - 4 459 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600007060.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000108.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the XM_005249033.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MYLIP

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          nervous system development
          negative regulation of neuron projection development
          negative regulation of low-density lipoprotein particle clearance
          protein ubiquitination
          protein destabilization
          regulation of low-density lipoprotein particle receptor catabolic process
          cholesterol homeostasis
          positive regulation of protein catabolic process
          ubiquitin-protein transferase activity
          protein binding
          cytoskeletal protein binding
          zinc ion binding
          transferase activity
          metal ion binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06001115.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06001115.hg.1 ENST00000356840.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06001115.hg.1 NM_013262.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06001115.hg.1 TC06000108.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 ENST00000584938.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007060.hg.1 NR_039782.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 ENST00000584938.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay TC0600007060.hg.1 NR_039782.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600148196.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600148196.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600148196.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600148196.hg.1 ENST00000349606.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0600148196.hg.1 ENST00000356840.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600148196.hg.1 NM_013262.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600148196.hg.1 TC0600007060.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline