Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00989424_m1
          See other MIER2 GE Assays ›
          Gene Symbol:
          MIER2
          Gene Name:
          MIER family member 2
          Gene Aliases:
          KIAA1193, Mi-er2
          Chromosome Location:
          Chr.19: 305573 - 344796 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00989424_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIER2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001346105.2 NP_001333034.1 11 - 12 1123 59
          NM_001387152.1 NP_001374081.1 12 - 13 1856 59
          NM_001387153.1 NP_001374082.1 12 - 13 1193 59
          NM_001387154.1 NP_001374083.1 11 - 12 1102 59
          NM_001387155.1 NP_001374084.1 12 - 13 2381 59
          NM_001387156.1 NP_001374085.1 13 - 14 1331 59
          NM_001387157.1 NP_001374086.1 12 - 13 1240 59
          NM_017550.3 NP_060020.1 12 - 13 1214 59
          XM_006722769.5 XP_006722832.1 12 - 13 1835 59
          XM_047438969.1 XP_047294925.1 12 - 13 1856 59
          XM_047438970.1 XP_047294926.1 12 - 13 1214 59
          XM_047438971.1 XP_047294927.1 12 - 13 1835 59
          XM_047438972.1 XP_047294928.1 11 - 12 1123 59
          XM_047438973.1 XP_047294929.1 12 - 13 4917 59
          XM_047438974.1 XP_047294930.1 13 - 14 3987 59
          XM_047438975.1 XP_047294931.1 13 - 14 2475 59
          XM_047438976.1 XP_047294932.1 12 - 13 2381 59
          XM_047438977.1 XP_047294933.1 13 - 14 1950 59
          XM_047438978.1 XP_047294934.1 13 - 14 1308 59
          XM_047438979.1 XP_047294935.1 10 - 11 984 59
          GenBank mRNA AB033019.1 - 1197 59
          BC028203.1 - 12 - 13 1209 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900009078.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000982.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MIER2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription corepressor activity
          histone deacetylase activity
          protein binding
          histone deacetylase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19013385.hg.1 ENST00000264819.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19013385.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19013385.hg.1 TC19000982.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000587966.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000587966.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000592722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000592722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000592722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000592722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000592722.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000619835.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000619835.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000619835.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000619835.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000621180.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000621180.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000621180.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000621180.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 ENST00000621180.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19013386.hg.1 ENST00000264819.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19013386.hg.1 NM_017550.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19013386.hg.1 TC19000982.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1900172040.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900172040.hg.1 ENST00000619835.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900172040.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900172040.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900172040.hg.1 ENST00000619835.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900172040.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900172040.hg.1 TC1900009078.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000586994.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000587966.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000587966.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000592722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000592722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000592722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000592722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000592722.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000619835.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000619835.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000619835.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000619835.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000621180.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000621180.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000621180.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000621180.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 ENST00000621180.4 Ensembl
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay TC1900009078.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900172039.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900172039.hg.1 ENST00000619835.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900172039.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900172039.hg.1 ENST00000264819.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900172039.hg.1 ENST00000619835.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1900172039.hg.1 NM_017550.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900172039.hg.1 TC1900009078.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline