Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00990130_m1
          See other GUCY2C GE Assays ›
          Gene Symbol:
          GUCY2C
          Gene Name:
          guanylate cyclase 2C
          Gene Aliases:
          DIAR6, GUC2C, MECIL, MUCIL, STAR
          Chromosome Location:
          Chr.12: 14612632 - 14696625 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00990130_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GUCY2C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004963.3 NP_004954.2 9 - 10 1305 151
          XM_011520631.2 XP_011518933.1 9 - 10 1203 151
          GenBank mRNA BC136544.1 - 9 - 10 1214 9020534 151
          BC136545.1 - 9 - 10 1214 9020535 151
          M73489.1 - 9 - 10 1216 151
          S57551.1 - 9 - 10 1285 151

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200010009.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001271.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GUCY2C

          Panther Classification:

          Molecular Function -

          guanylate cyclase

          Gene Ontology Categories:

          Function(s) Process(es)

          cGMP biosynthetic process
          protein phosphorylation
          receptor guanylyl cyclase signaling pathway
          response to toxic substance
          intracellular signal transduction
          regulation of cell proliferation
          adenylate cyclase activity
          guanylate cyclase activity
          protein kinase activity
          protein binding
          ATP binding
          GTP binding
          toxic substance binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12016551.hg.1 ENST00000261170.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12016551.hg.1 NM_004963.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12016551.hg.1 TC12001271.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200175573.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200175573.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200175573.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200175573.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200175573.hg.1 TC1200010009.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000261170.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 ENST00000535803.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq
          Affymetrix Human Clariom D Assay TC1200010009.hg.1 NM_004963.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-rd95r:80/100.66.77.199:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline