Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01026335_m1
          See other SV2C GE Assays ›
          Gene Symbol:
          SV2C ADAM12
          Gene Name:
          synaptic vesicle glycoprotein 2C
          ADAM metallopeptidase domain 12
          Gene Aliases:
          SLC22B3
          ADAM12-OT1, CAR10, MCMP, MCMPMltna, MLTN, MLTNA
          Chromosome Location:
          Chr.5: 75847464 - 76353939 on Build GRCh38
          Chromosome Location:
          Chr.10: 126012391 - 126388477 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01026335_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SV2C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001297716.2 NP_001284645.1 8 - 9 1567 101
          NM_014979.4 NP_055794.3 8 - 9 1567 101
          XM_011543281.4 XP_011541583.1 8 - 9 1535 101
          XM_011543282.4 XP_011541584.2 8 - 9 2462 101
          XM_017009244.3 XP_016864733.1 4 - 5 444 101
          GenBank mRNA AB028977.1 - 6 - 7 597 101
          AK094917.1 - 8 - 9 1537 101
          BC100824.1 - 8 - 9 1472 101
          BC100825.1 - 8 - 9 1470 101
          BC100826.1 - 8 - 9 1470 101
          BC100827.1 - 8 - 9 1470 101
          Gene Symbol: ADAM12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF023477.2 - 8 - 9 1470 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500007820.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000366.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SV2C
          Click here to find siRNAs for Gene ADAM12

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter transport
          chemical synaptic transmission
          transmembrane transport
          regulation of synaptic vesicle exocytosis
          protein binding
          transmembrane transporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000506257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000506257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000506257.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 ENST00000581282.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05005117.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05005117.hg.1 ENST00000502798.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05005117.hg.1 NM_014979.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05005117.hg.1 TC05000366.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000506257.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000506257.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000506257.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 ENST00000581282.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay TC0500007820.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500153475.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500153475.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500153475.hg.1 ENST00000506257.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500153475.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500153475.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500153475.hg.1 ENST00000322285.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0500153475.hg.1 ENST00000502798.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0500153475.hg.1 ENST00000506257.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500153475.hg.1 NM_001297716.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500153475.hg.1 NM_014979.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500153475.hg.1 TC0500007820.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline