| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000483769.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000483769.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000483769.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000497568.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000497568.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000497568.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
ENST00000497568.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02039073.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02039073.hg.1 |
ENST00000418842.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02039073.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02039073.hg.1 |
TC02002479.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02039074.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02039074.hg.1 |
ENST00000418842.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02039074.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02039074.hg.1 |
TC02002479.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000483769.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000483769.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000483769.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000497568.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000497568.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000497568.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
ENST00000497568.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014770.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200207515.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207515.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207515.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207515.hg.1 |
ENST00000497568.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207515.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200207515.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207515.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207515.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207515.hg.1 |
ENST00000497568.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207515.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200207515.hg.1 |
TC0200014770.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200207516.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207516.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207516.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207516.hg.1 |
ENST00000497568.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207516.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200207516.hg.1 |
ENST00000375497.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207516.hg.1 |
ENST00000418842.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207516.hg.1 |
ENST00000492913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207516.hg.1 |
ENST00000497568.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200207516.hg.1 |
NM_002054.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200207516.hg.1 |
TC0200014770.hg.1 |
Clariom D TC |