Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01034270_m1
          See other GTF3C4 GE Assays ›
          Gene Symbol:
          GTF3C4
          Gene Name:
          general transcription factor IIIC subunit 4
          Gene Aliases:
          KAT12, TF3C-delta, TFIII90, TFIIIC290, TFIIIC90, TFIIICDELTA
          Chromosome Location:
          Chr.9: 132670035 - 132694955 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01034270_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GTF3C4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012204.4 NP_036336.2 1 - 2 486 75
          NR_133925.1 1 - 2 915 75
          GenBank mRNA AF142328.1 - 1 - 2 603 75
          BC011619.2 - 1 - 2 439 75
          BC022787.1 - 1 - 2 439 75
          BC094774.1 - 1 - 2 460 75
          BC104755.1 - 1 - 2 420 75
          BC112245.1 - 1 - 2 420 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900009032.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000760.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GTF3C4

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin remodeling
          transcription from RNA polymerase III promoter
          transcription initiation from RNA polymerase III promoter
          5S class rRNA transcription from RNA polymerase III type 1 promoter
          tRNA transcription from RNA polymerase III promoter
          transcription factor activity, core RNA polymerase III binding
          DNA binding
          histone acetyltransferase activity
          protein binding
          transferase activity
          transferase activity, transferring acyl groups
          histone acetyltransferase activity (H3-K14 specific)
          peptide-lysine-N-acetyltransferase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09009756.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09009756.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09009756.hg.1 TC09000760.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09009757.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09009757.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09009757.hg.1 TC09000760.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09009758.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09009758.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09009758.hg.1 TC09000760.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09009760.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09009760.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09009760.hg.1 TC09000760.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR09009755.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09009755.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09009755.hg.1 TC09000760.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09009759.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09009759.hg.1 NM_012204.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09009759.hg.1 TC09000760.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay TC0900009032.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161444.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161444.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161444.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161444.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161444.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161444.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161444.hg.1 TC0900009032.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900161445.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161445.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161445.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161445.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161445.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161445.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161445.hg.1 TC0900009032.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900161448.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161448.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161448.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161448.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161448.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161448.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161448.hg.1 TC0900009032.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900161442.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161442.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161442.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161442.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161442.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161442.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161442.hg.1 TC0900009032.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900161443.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161443.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161443.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161443.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161443.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161443.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161443.hg.1 TC0900009032.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900161447.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161447.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161447.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161447.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161447.hg.1 TC0900009032.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900161446.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161446.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161446.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161446.hg.1 ENST00000372146.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161446.hg.1 ENST00000483873.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900161446.hg.1 NM_012204.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900161446.hg.1 TC0900009032.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 154619 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline