Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01058182_m1
          See other C5ORF66-AS2 GE Assays ›
          Gene Symbol:
          C5orf66-AS2
          Gene Name:
          C5orf66 antisense RNA 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 135236234 - 135248177 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01058182_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C5orf66-AS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037895.1 3 - 4 406 72
          GenBank mRNA BC045187.1 - 3 - 4 408 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500012098.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05001800.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299048 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299007 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299065 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299046 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299023 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299025 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299099 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477199 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299035 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15742995 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299032 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298994 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298976 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15742998 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299003 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298998 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477193 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299084 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299000 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298990 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299086 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299098 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298977 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299055 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299064 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299091 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298991 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298996 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299031 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299016 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299105 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299077 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299075 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15742997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477102 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477138 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298983 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299094 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299066 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477104 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299079 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299080 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298992 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299067 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298995 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299090 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298988 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299083 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299012 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477134 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299068 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298969 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298975 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299088 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299070 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299005 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477212 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299051 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15742999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299093 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299061 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299021 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299050 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04299010 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04298985 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24477182 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3660204 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P117804 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 32240 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 14414 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3660204 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3660204 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P117804 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P117804 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05024468.hg.1 TC05001800.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05024468.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024468.hg.1 NR_037895.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024469.hg.1 TC05001800.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05024469.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024469.hg.1 NR_037895.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024473.hg.1 TC05001800.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05024473.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024473.hg.1 NR_037895.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012098.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012098.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012098.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012098.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012098.hg.1 ENST00000555344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012098.hg.1 ENST00000555344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500012098.hg.1 NR_037895.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012098.hg.1 NR_037895.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012098.hg.1 NR_037895.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500012098.hg.1 NR_037895.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05024471.hg.1 TC05001800.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05024471.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024471.hg.1 ENST00000555344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05024471.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay TC0500012098.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500012098.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500012098.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500012098.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500012098.hg.1 ENST00000555344.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500012098.hg.1 ENST00000555344.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500012098.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay TC0500012098.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay TC0500012098.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay TC0500012098.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183697.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183697.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183697.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183697.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183697.hg.1 TC0500012098.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500183699.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183699.hg.1 ENST00000555344.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183699.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183699.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183699.hg.1 ENST00000555344.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183699.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183699.hg.1 TC0500012098.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500183701.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183701.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183701.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183701.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183701.hg.1 TC0500012098.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500183696.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183696.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183696.hg.1 ENST00000505663.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500183696.hg.1 NR_037895.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500183696.hg.1 TC0500012098.hg.1 Clariom D TC
          Human Genome U95 Set 65012_PM_at - -
          Human Genome U95 Set 65012_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A027424 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A027424 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1915805 BC045187.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1915805 NR_037895.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1915805 BC045187.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1915805 NR_037895.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1915805 BC045187.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1915805 NR_037895.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 6560441 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_91216 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1915805 BC045187.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1915805 NR_037895.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1915805 BC045187.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1915805 NR_037895.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1915805 BC045187.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1915805 NR_037895.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline