Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01072500_s1
          See other LOC728024 GE Assays ›
          Gene Symbol:
          LOC728024
          Gene Name:
          chromosome X open reading frame 56 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 37746554 - 37748046 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01072500_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC728024
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003671.2 1 1376 63
          GenBank mRNA BM679328.1 - 1 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800010133.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08001136.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P171223 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P290373 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358222 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076257 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076200 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076196 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076251 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076203 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358176 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076235 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358221 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358199 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16087326 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076193 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358208 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076204 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076273 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16087319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16087328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358225 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16087325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358200 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076207 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16087317 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076215 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16087329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076260 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076194 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076213 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358230 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358176 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358155 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358152 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358174 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076194 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16087328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076251 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358164 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076200 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358199 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076236 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358221 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076227 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076257 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076235 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16087317 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076193 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076273 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358225 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358222 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076207 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076215 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16087319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358208 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16087329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358166 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076198 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076231 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076244 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076204 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076260 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16087326 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358200 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076203 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076238 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16087325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076196 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P06076233 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358157 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26358165 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076268 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076132 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076254 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076141 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076263 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076202 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076146 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076154 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076150 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076135 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076235 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076221 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076207 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076232 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076277 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076201 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076209 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076196 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16087328 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076194 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16087319 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076215 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076241 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076244 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076139 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076247 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076156 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076159 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076199 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076203 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076217 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076152 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076237 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076260 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076252 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076258 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076274 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076144 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076212 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076249 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076270 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076239 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076275 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16087302 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16087322 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076272 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076224 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16087329 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076279 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16087317 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076138 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076219 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076227 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P06076245 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_003671_at - -
          Affymetrix Human Gene 1.0 ST Array 8150218 - -
          Affymetrix Human Gene 1.1 ST Array 8150217 - -
          Affymetrix Human Transcriptome Array 2.0 PSR08014559.hg.1 TC08001136.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08014559.hg.1 NR_003671.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08014561.hg.1 TC08001136.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08014561.hg.1 NR_003671.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08014560.hg.1 TC08001136.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08014560.hg.1 ENST00000453751.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08014560.hg.1 NR_003671.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800168873.hg.1 ENST00000521192.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800168873.hg.1 NR_003671.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800168873.hg.1 ENST00000521192.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0800168873.hg.1 NR_003671.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800168873.hg.1 TC0800010133.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800168874.hg.1 NR_003671.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800168874.hg.1 NR_003671.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800168874.hg.1 TC0800010133.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800168872.hg.1 NR_003671.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800168872.hg.1 NR_003671.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800168872.hg.1 TC0800010133.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035686 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035686 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 141214 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3235036 BI758198.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3235036 CR977513.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3235036 NR_003671.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3235036 BI758198.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3235036 CR977513.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3235036 NR_003671.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3235036 BI758198.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3235036 CR977513.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3235036 NR_003671.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3235036 BI758198.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3235036 CR977513.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3235036 NR_003671.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-qjfm9:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline