Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01089718_m1
          See other CCDC130 GE Assays ›
          Gene Symbol:
          CCDC130
          Gene Name:
          coiled-coil domain containing 130
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 13731760 - 13763298 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01089718_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCDC130
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001320561.1 NP_001307490.1 7 - 8 889 66
          NM_001320564.1 NP_001307493.1 7 - 8 877 66
          NM_001320565.1 NP_001307494.1 7 - 8 759 66
          NM_001320566.1 NP_001307495.1 7 - 8 877 66
          NM_001320567.1 NP_001307496.1 7 - 8 740 66
          NM_001320568.1 NP_001307497.1 6 - 7 549 66
          NM_001320569.1 NP_001307498.1 5 - 6 345 66
          NM_030818.3 NP_110445.1 6 - 7 549 66
          XM_005260086.4 XP_005260143.1 7 - 8 739 66
          XM_011528326.1 XP_011526628.1 6 - 7 628 66
          GenBank mRNA AF250306.1 - 6 - 7 477 66
          AK098262.1 - 6 - 7 506 66
          AK293927.1 - 7 - 8 740 66
          AL560878.3 - 7 - 8 760 66
          BC002905.2 - 6 - 7 473 3940130 66
          BI913255.1 - 5 - 6 310 66
          BM547551.1 - 6 - 7 547 66
          BX394050.2 - 7 - 8 870 66
          BX422326.2 - 6 - 7 548 66
          CR999732.1 - 5 - 6 474 66

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001320566.1, NM_001320567.1, NM_001320568.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CCDC130

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to virus
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000587019.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000587019.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000587019.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000587019.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000587019.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000589096.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000589096.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000589096.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000589096.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000589096.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000589669.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000589669.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000589669.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000589669.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000589669.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000589669.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000221554.12 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000585844.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000586666.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000587019.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000587019.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000587019.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000587019.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000587019.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000588809.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000589096.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000589096.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000589096.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000589096.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000589096.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000589669.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000589669.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000589669.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000589669.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000589669.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000589669.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 ENST00000593174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011673.hg.1 NM_030818.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline