Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01104808_m1
          See other MAP4 GE Assays ›
          Gene Symbol:
          MAP4
          Gene Name:
          microtubule associated protein 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 47850695 - 48088848 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01104808_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAP4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134364.2 NP_001127836.1 8 - 9 2094 93
          NM_001384675.1 NP_001371604.1 7 - 8 1971 93
          NM_001384676.1 NP_001371605.1 8 - 9 2094 93
          NM_001384677.1 NP_001371606.1 8 - 9 2094 93
          NM_001384678.1 NP_001371607.1 9 - 10 2130 93
          NM_001384680.1 NP_001371609.1 5 - 6 1788 93
          NM_001384681.1 NP_001371610.1 7 - 8 1971 93
          NM_001384707.1 NP_001371636.1 7 - 8 1971 93
          NM_001384729.1 NP_001371658.1 10 - 11 2299 93
          NM_001384730.1 NP_001371659.1 7 - 8 2025 93
          NM_001384731.1 NP_001371660.1 9 - 10 2145 93
          NM_001384733.1 NP_001371662.1 8 - 9 2094 93
          NM_001384734.1 NP_001371663.1 8 - 9 2094 93
          NM_001384735.1 NP_001371664.1 8 - 9 2094 93
          NM_001384736.1 NP_001371665.1 9 - 10 2145 93
          NM_001384737.1 NP_001371666.1 8 - 9 2094 93
          NM_001384738.1 NP_001371667.1 9 - 10 2130 93
          NM_001384744.1 NP_001371673.1 8 - 9 2022 93
          NM_001384746.1 NP_001371675.1 6 - 7 1902 93
          NM_001384748.1 NP_001371677.1 9 - 10 2130 93
          NM_001384751.1 NP_001371680.1 8 - 9 2094 93
          NM_001384752.1 NP_001371681.1 8 - 9 2094 93
          NM_001384753.1 NP_001371682.1 6 - 7 1902 93
          NM_001384754.1 NP_001371683.1 8 - 9 2094 93
          NM_001384755.1 NP_001371684.1 7 - 8 1971 93
          NM_001384756.1 NP_001371685.1 8 - 9 2094 93
          NM_001384757.1 NP_001371686.1 6 - 7 1902 93
          NM_001384758.1 NP_001371687.1 7 - 8 1980 93
          NM_001384759.1 NP_001371688.1 7 - 8 1971 93
          NM_001384760.1 NP_001371689.1 7 - 8 1971 93
          NM_001384761.1 NP_001371690.1 8 - 9 2094 93
          NM_001384762.1 NP_001371691.1 7 - 8 870 93
          NM_001384774.1 NP_001371703.1 8 - 9 2094 93
          NM_001384776.1 NP_001371705.1 7 - 8 1980 93
          NM_001384777.1 NP_001371706.1 7 - 8 2025 93
          NM_001384778.1 NP_001371707.1 8 - 9 1461 93
          NM_001384779.1 NP_001371708.1 8 - 9 2094 93
          NM_001384780.1 NP_001371709.1 7 - 8 2025 93
          NM_001384781.1 NP_001371710.1 8 - 9 2094 93
          NM_001384782.1 NP_001371711.1 7 - 8 2025 93
          NM_001384783.1 NP_001371712.1 7 - 8 2025 93
          NM_001384784.1 NP_001371713.1 7 - 8 870 93
          NM_001384785.1 NP_001371714.1 8 - 9 2094 93
          NM_001384786.1 NP_001371715.1 7 - 8 2025 93
          NM_001384787.1 NP_001371716.1 7 - 8 2025 93
          NM_001384788.1 NP_001371717.1 9 - 10 2145 93
          NM_001384789.1 NP_001371718.1 8 - 9 2022 93
          NM_001384790.1 NP_001371719.1 9 - 10 2145 93
          NM_001384791.1 NP_001371720.1 8 - 9 2022 93
          NM_001384792.1 NP_001371721.1 9 - 10 2145 93
          NM_001384793.1 NP_001371722.1 8 - 9 2022 93
          NM_001384794.1 NP_001371723.1 9 - 10 2145 93
          NM_001384795.1 NP_001371724.1 10 - 11 2181 93
          NM_001384796.1 NP_001371725.1 9 - 10 2145 93
          NM_001384797.1 NP_001371726.1 8 - 9 2022 93
          NM_001384798.1 NP_001371727.1 8 - 9 2094 93
          NM_001384800.1 NP_001371729.1 8 - 9 2094 93
          NM_001384803.1 NP_001371732.1 6 - 7 1857 93
          NM_001384804.1 NP_001371733.1 7 - 8 1971 93
          NM_001384805.1 NP_001371734.1 7 - 8 1971 93
          NM_001384806.1 NP_001371735.1 7 - 8 1971 93
          NM_001384807.1 NP_001371736.1 6 - 7 1857 93
          NM_001384809.1 NP_001371738.1 8 - 9 2094 93
          NM_001384810.1 NP_001371739.1 8 - 9 2094 93
          NM_001384811.1 NP_001371740.1 7 - 8 2025 93
          NM_001384813.1 NP_001371742.1 7 - 8 792 93
          NM_001384814.1 NP_001371743.1 8 - 9 2094 93
          NM_001384816.1 NP_001371745.1 7 - 8 2025 93
          NM_001384819.1 NP_001371748.1 8 - 9 2094 93
          NM_001384820.1 NP_001371749.1 8 - 9 2094 93
          NM_001384824.1 NP_001371753.1 8 - 9 2094 93
          NM_001384825.1 NP_001371754.1 7 - 8 1980 93
          NM_001384826.1 NP_001371755.1 7 - 8 1971 93
          NM_001384827.1 NP_001371756.1 8 - 9 2094 93
          NM_001384828.1 NP_001371757.1 7 - 8 2025 93
          NM_001384831.1 NP_001371760.1 8 - 9 2094 93
          NM_001384832.1 NP_001371761.1 7 - 8 1980 93
          NM_001384834.1 NP_001371763.1 8 - 9 2004 93
          NM_001384836.1 NP_001371765.1 7 - 8 1971 93
          NM_001384837.1 NP_001371766.1 7 - 8 1971 93
          NM_001384838.1 NP_001371767.1 8 - 9 2094 93
          NM_001384839.1 NP_001371768.1 7 - 8 1852 93
          NM_001384840.1 NP_001371769.1 8 - 9 2094 93
          NM_001384841.1 NP_001371770.1 7 - 8 1980 93
          NM_001384842.1 NP_001371771.1 6 - 7 1857 93
          NM_001384843.1 NP_001371772.1 6 - 7 1857 93
          NM_001384844.1 NP_001371773.1 8 - 9 2094 93
          NM_001384845.1 NP_001371774.1 7 - 8 1852 93
          NM_001384846.1 NP_001371775.1 8 - 9 2094 93
          NM_001384847.1 NP_001371776.1 8 - 9 2094 93
          NM_001384848.1 NP_001371777.1 8 - 9 2090 93
          NM_001384849.1 NP_001371778.1 8 - 9 2055 93
          NM_001384850.1 NP_001371779.1 7 - 8 1971 93
          NM_001384851.1 NP_001371780.1 7 - 8 1980 93
          NM_001384853.1 NP_001371782.1 7 - 8 1971 93
          NM_001384856.1 NP_001371785.1 8 - 9 1461 93
          NM_001384857.1 NP_001371786.1 7 - 8 2025 93
          NM_001384859.1 NP_001371788.1 8 - 9 2016 93
          NM_001384861.1 NP_001371790.1 8 - 9 2094 93
          NM_001384862.1 NP_001371791.1 6 - 7 1729 93
          NM_001384863.1 NP_001371792.1 9 - 10 2178 93
          NM_001384864.1 NP_001371793.1 6 - 7 1902 93
          NM_001384866.1 NP_001371795.1 7 - 8 1971 93
          NM_001384867.1 NP_001371796.1 7 - 8 1852 93
          NM_001384868.1 NP_001371797.1 7 - 8 1971 93
          NM_001384869.1 NP_001371798.1 7 - 8 2025 93
          NM_001384870.1 NP_001371799.1 8 - 9 2094 93
          NM_001384871.1 NP_001371800.1 8 - 9 2094 93
          NM_001384872.1 NP_001371801.1 9 - 10 2130 93
          NM_001384873.1 NP_001371802.1 8 - 9 2016 93
          NM_001384874.1 NP_001371803.1 8 - 9 2004 93
          NM_001384875.1 NP_001371804.1 9 - 10 2130 93
          NM_001384876.1 NP_001371805.1 7 - 8 1980 93
          NM_001384877.1 NP_001371806.1 6 - 7 747 93
          NM_001384879.1 NP_001371808.1 8 - 9 2094 93
          NM_001384892.1 NP_001371821.1 8 - 9 2004 93
          NM_001384893.1 NP_001371822.1 7 - 8 1971 93
          NM_001385665.1 NP_001372594.1 8 - 9 2101 93
          NM_001385675.1 NP_001372604.1 9 - 10 2137 93
          NM_001385676.1 NP_001372605.1 6 - 7 1909 93
          NM_001385677.1 NP_001372606.1 8 - 9 2101 93
          NM_001385681.1 NP_001372610.1 9 - 10 2152 93
          NM_001385684.1 NP_001372613.1 8 - 9 2101 93
          NM_001385685.1 NP_001372614.1 8 - 9 2101 93
          NM_001385686.1 NP_001372615.1 9 - 10 2152 93
          NM_002375.5 NP_002366.2 8 - 9 2094 93
          XM_017006397.2 XP_016861886.1 10 - 11 2181 93
          XM_047448130.1 XP_047304086.1 10 - 11 2181 93
          XM_047448131.1 XP_047304087.1 10 - 11 2181 93
          XM_047448132.1 XP_047304088.1 8 - 9 2022 93
          XM_047448133.1 XP_047304089.1 7 - 8 1971 93
          GenBank mRNA AB209377.1 - 8 - 9 2085 93
          AK297242.1 - 7 - 8 2024 93
          BG760540.1 - 2 - 3 221 93
          M64571.1 - 8 - 9 2089 93
          U19727.1 - 8 - 9 2074 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300010999.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001372.hg.1

          Set Membership:

          Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          mitotic spindle organization
          neuron projection development
          microtubule polymerization
          microtubule sliding
          establishment of spindle orientation
          cell division
          cilium disassembly
          negative regulation of nonmotile primary cilium assembly
          RNA binding
          structural molecule activity
          protein binding
          microtubule binding
          tubulin binding
          microtubule stabilizing activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03023680.hg.1 ENST00000264724.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023680.hg.1 ENST00000360240.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023680.hg.1 ENST00000383737.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023680.hg.1 ENST00000395734.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023680.hg.1 ENST00000420772.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023680.hg.1 ENST00000426837.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023680.hg.1 ENST00000441748.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023680.hg.1 ENST00000462206.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03023680.hg.1 NM_001134364.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03023680.hg.1 NM_002375.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03023680.hg.1 TC03001372.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000335271.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000360240.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000383736.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000395734.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000420772.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000426837.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000429422.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000462206.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000482752.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000497735.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 NM_001134364.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 NM_002375.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000335271.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000360240.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000383736.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000395734.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000420772.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000426837.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000429422.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000462206.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000482752.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 ENST00000497735.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 NM_001134364.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 NM_002375.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300180425.hg.1 TC0300010999.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline