Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01120908_m1
          See other SDC4 GE Assays ›
          Gene Symbol:
          SDC4
          Gene Name:
          syndecan 4
          Gene Aliases:
          SYND4
          Chromosome Location:
          Chr.20: 45325288 - 45348424 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01120908_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SDC4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002999.3 NP_002990.2 3 - 4 285 147
          XM_011528977.2 XP_011527279.1 2 - 3 140 147
          GenBank mRNA AK222695.1 - 3 - 4 285 147
          AK223243.1 - 3 - 4 287 147
          AK303964.1 - 2 - 3 146 147
          AK312507.1 - 3 - 4 285 147
          BC030805.1 - 3 - 4 285 4690309 147
          CR542045.1 - 3 - 4 245 147
          CR542074.1 - 3 - 4 245 147
          D13292.1 - 3 - 4 285 147
          X67016.1 - 3 - 4 271 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000009218.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000878.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK303964.1, XM_011528977.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SDC4

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          ureteric bud development
          neural tube closure
          glycosaminoglycan biosynthetic process
          glycosaminoglycan catabolic process
          signal transduction
          regulation of fibroblast migration
          cell migration
          glycosaminoglycan metabolic process
          wound healing
          positive regulation of protein kinase activity
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          inner ear receptor stereocilium organization
          positive regulation of exosomal secretion
          positive regulation of extracellular exosome assembly
          fibronectin binding
          protein kinase C binding
          protein binding
          thrombospondin receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR20011982.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20011982.hg.1 ENST00000537976.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20011982.hg.1 NM_002999.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20011982.hg.1 TC20000878.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20011983.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20011983.hg.1 ENST00000537976.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20011983.hg.1 NM_002999.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20011983.hg.1 TC20000878.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Clariom D Assay TC2000009218.hg.1 NM_002999.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000164492.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2000164492.hg.1 NM_002999.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000164492.hg.1 TC2000009218.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000164492.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2000164492.hg.1 NM_002999.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000164491.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2000164491.hg.1 NM_002999.3 Refseq
          Affymetrix Human Clariom D Assay PSR2000164491.hg.1 TC2000009218.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000164491.hg.1 ENST00000372733.3 Ensembl
          Affymetrix Human Clariom D Assay PSR2000164491.hg.1 NM_002999.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline