Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01124134_m1
          See other CYP17A1 GE Assays ›
          Gene Symbol:
          CYP17A1
          Gene Name:
          cytochrome P450 family 17 subfamily A member 1
          Gene Aliases:
          CPT7, CYP17, P450C17, S17AH
          Chromosome Location:
          Chr.10: 102830531 - 102837413 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01124134_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CYP17A1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000102.4 NP_000093.1 4 - 5 808 120
          GenBank mRNA AK289898.1 - 4 - 5 808 120
          AK308466.1 - 4 - 5 718 120
          AK312235.1 - 4 - 5 810 120
          BC062997.1 - 4 - 5 780 120
          BC063388.1 - 4 - 5 780 120
          BT020000.1 - 4 - 5 756 120
          M14564.1 - 4 - 5 797 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000011696.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001625.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection P450 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CYP17A1

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          steroid biosynthetic process
          androgen biosynthetic process
          glucocorticoid biosynthetic process
          sex differentiation
          steroid metabolic process
          cortisol biosynthetic process
          hormone metabolic process
          hormone biosynthetic process
          progesterone metabolic process
          olefinic compound metabolic process
          monooxygenase activity
          steroid 17-alpha-monooxygenase activity
          iron ion binding
          steroid hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          lyase activity
          oxygen binding
          heme binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10021193.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021193.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10021193.hg.1 TC10001625.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000469683.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000469683.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000469683.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10021194.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10021194.hg.1 NM_000102.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10021194.hg.1 TC10001625.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000469683.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000469683.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000469683.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 ENST00000489268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay TC1000011696.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000180060.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000180060.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000180060.hg.1 ENST00000369887.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1000180060.hg.1 NM_000102.3 Refseq
          Affymetrix Human Clariom D Assay PSR1000180060.hg.1 TC1000011696.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0